missing translation for 'onlineSavingsMsg'
Learn More

4-[(4-bromothien-2-yl)methyl]morpholine, 97%, Thermo Scientific™

275.00 € - 354.00 €

Chemical Identifiers

CAS 194851-19-9
Molecular Formula C9H12BrNOS
Molecular Weight (g/mol) 262.17
MDL Number MFCD01653482
InChI Key XYQVGDMXJNORKK-UHFFFAOYSA-N
Synonym 4-4-bromothiophen-2-yl methyl morpholine, 4-4-bromothien-2-yl methyl morpholine, 4-bromo-2-morpholin-4-ylmethyl thiophene, cambridge id 5430345, 4-bromo-2-morpholinomethylthiophene, 4-4-bromo-thiophen-2-ylmethyl-morpholine, morpholine, 4-4-bromo-2-thienyl methyl
PubChem CID 783497
IUPAC Name 4-[(4-bromothiophen-2-yl)methyl]morpholine
SMILES BrC1=CSC(CN2CCOCC2)=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10525944
View Documents Promotion Details
Thermo Scientific Maybridge
CC74210DA
1 g Amber Glass Bottle
275.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10535944
View Documents Promotion Details
Thermo Scientific Maybridge
CC74210DE
5 g Amber Glass Bottle
354.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

194851-19-9
262.17
XYQVGDMXJNORKK-UHFFFAOYSA-N
783497
BrC1=CSC(CN2CCOCC2)=C1
C9H12BrNOS
MFCD01653482
4-4-bromothiophen-2-yl methyl morpholine, 4-4-bromothien-2-yl methyl morpholine, 4-bromo-2-morpholin-4-ylmethyl thiophene, cambridge id 5430345, 4-bromo-2-morpholinomethylthiophene, 4-4-bromo-thiophen-2-ylmethyl-morpholine, morpholine, 4-4-bromo-2-thienyl methyl
4-[(4-bromothiophen-2-yl)methyl]morpholine

Specifications

194851-19-9
100.0
C9H12BrNOS
4-4-bromothiophen-2-yl methyl morpholine, 4-4-bromothien-2-yl methyl morpholine, 4-bromo-2-morpholin-4-ylmethyl thiophene, cambridge id 5430345, 4-bromo-2-morpholinomethylthiophene, 4-4-bromo-thiophen-2-ylmethyl-morpholine, morpholine, 4-4-bromo-2-thienyl methyl
BrC1=CSC(CN2CCOCC2)=C1
262.17
262.17
4-[(4-Bromothien-2-yl)methyl]morpholine
96.5
97%
MFCD01653482
XYQVGDMXJNORKK-UHFFFAOYSA-N
4-[(4-bromothiophen-2-yl)methyl]morpholine
783497
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications