Learn More
4-(4-Bromophenyl)butanoic acid, 97%, Thermo Scientific™
53.10 € - 311.00 €
Chemical Identifiers
CAS | 35656-89-4 |
---|---|
Molecular Formula | C10H11BrO2 |
Molecular Weight (g/mol) | 243.1 |
MDL Number | MFCD00463170 |
InChI Key | AGIIMNQWNPUJPT-UHFFFAOYSA-N |
Synonym | 4-4-bromophenyl butanoic acid, 4-4-bromophenyl butyric acid, benzenebutanoic acid, 4-bromo, 4-bromo-benzenebutanoic acid, pubchem16174, acmc-209iig, 4-bromobenzenebutanoic acid, benzenebutanoic acid,4-bromo, 4-4-bromo-phenyl-butyric acid |
PubChem CID | 252732 |
IUPAC Name | 4-(4-bromophenyl)butanoic acid |
SMILES | C1=CC(=CC=C1CCCC(=O)O)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10513983
|
Thermo Scientific Maybridge
MO01273CB |
250 mg |
53.10 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10747664
|
Thermo Scientific Maybridge
MO01273DA |
1 g |
105.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10651036
|
Thermo Scientific Maybridge
MO01273DE |
5 g |
311.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
35656-89-4 | |
243.1 | |
AGIIMNQWNPUJPT-UHFFFAOYSA-N | |
252732 | |
C1=CC(=CC=C1CCCC(=O)O)Br |
C10H11BrO2 | |
MFCD00463170 | |
4-4-bromophenyl butanoic acid, 4-4-bromophenyl butyric acid, benzenebutanoic acid, 4-bromo, 4-bromo-benzenebutanoic acid, pubchem16174, acmc-209iig, 4-bromobenzenebutanoic acid, benzenebutanoic acid,4-bromo, 4-4-bromo-phenyl-butyric acid | |
4-(4-bromophenyl)butanoic acid |
Specifications
35656-89-4 | |
97% | |
C10H11BrO2 | |
4-4-bromophenyl butanoic acid, 4-4-bromophenyl butyric acid, benzenebutanoic acid, 4-bromo, 4-bromo-benzenebutanoic acid, pubchem16174, acmc-209iig, 4-bromobenzenebutanoic acid, benzenebutanoic acid,4-bromo, 4-4-bromo-phenyl-butyric acid | |
C1=CC(=CC=C1CCCC(=O)O)Br | |
243.1 | |
243.1 | |
4-(4-Bromophenyl)butanoic acid |
97.0 | |
Amber glass bottle | |
MFCD00463170 | |
AGIIMNQWNPUJPT-UHFFFAOYSA-N | |
4-(4-bromophenyl)butanoic acid | |
252732 | |
97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.