Learn More
4,4',4″-Trimethyltriphenylamine, 98%, Thermo Scientific Chemicals
46.10 € - 583.00 €
Chemical Identifiers
CAS | 1159-53-1 |
---|---|
Molecular Formula | C21H21N |
Molecular Weight (g/mol) | 287.406 |
MDL Number | MFCD00674043 |
InChI Key | YXYUIABODWXVIK-UHFFFAOYSA-N |
Synonym | tri-p-tolylamine, 4,4',4-trimethyltriphenylamine, 4,4,4-trimethyltriphenylamine, tri p-tolyl amine, benzenamine, 4-methyl-n,n-bis 4-methylphenyl, 4-methyl-n,n-bis 4-methylphenyl aniline, tris 4-methylphenyl amine, n,n-di-p-tolyl-p-toluidine, n,n,n-tri p-tolyl amine, trip-tolylamine |
PubChem CID | 70873 |
IUPAC Name | 4-methyl-N,N-bis(4-methylphenyl)aniline |
SMILES | CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15411239
|
Thermo Scientific Alfa Aesar
H64162.03 |
1 g |
46.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15421239
|
Thermo Scientific Alfa Aesar
H64162.06 |
5 g |
156.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15431239
|
Thermo Scientific Alfa Aesar
H64162.14 |
25 g |
583.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used for proteomics research. Also employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed for proteomics research. Also employed as a intermediate for pharmaceutical.
Solubility
Soluble in chloroform and tetrahydrofuran (THF).
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
1159-53-1 | |
287.406 | |
YXYUIABODWXVIK-UHFFFAOYSA-N | |
70873 | |
CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C |
C21H21N | |
MFCD00674043 | |
tri-p-tolylamine, 4,4',4-trimethyltriphenylamine, 4,4,4-trimethyltriphenylamine, tri p-tolyl amine, benzenamine, 4-methyl-n,n-bis 4-methylphenyl, 4-methyl-n,n-bis 4-methylphenyl aniline, tris 4-methylphenyl amine, n,n-di-p-tolyl-p-toluidine, n,n,n-tri p-tolyl amine, trip-tolylamine | |
4-methyl-N,N-bis(4-methylphenyl)aniline |
Specifications
1159-53-1 | |
White | |
MFCD00674043 | |
tri-p-tolylamine, 4,4',4-trimethyltriphenylamine, 4,4,4-trimethyltriphenylamine, tri p-tolyl amine, benzenamine, 4-methyl-n,n-bis 4-methylphenyl, 4-methyl-n,n-bis 4-methylphenyl aniline, tris 4-methylphenyl amine, n,n-di-p-tolyl-p-toluidine, n,n,n-tri p-tolyl amine, trip-tolylamine | |
YXYUIABODWXVIK-UHFFFAOYSA-N | |
4-methyl-N,N-bis(4-methylphenyl)aniline | |
70873 | |
98% | |
4,4',4''-Trimethyltriphenylamine |
114°C to 118°C (literature) | |
C21H21N | |
2215309 | |
Soluble in chloroform and tetrahydrofuran (THF). | |
CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C | |
287.406 | |
287.41 | |
Solid |
Safety and Handling
missing translation for 'einecsNumber' : 214-595-1
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.