missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Piperidinyloxy)benzonitrile, 97%, Thermo Scientific™

Product Code. 11409780
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902837-34-7
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.257
MDL Number MFCD08061105
InChI Key KMBKWCFPFFJUPE-UHFFFAOYSA-N
Synonym 4-3-piperidinyloxy benzonitrile, 4-piperidin-3-yloxy benzonitrile, 3-4-cyanophenoxy piperidine hcl, 4-piperidin-3-yl oxy benzonitrile
PubChem CID 44721293
IUPAC Name 4-piperidin-3-yloxybenzonitrile
SMILES C1CC(CNC1)OC2=CC=C(C=C2)C#N

Specifications

Quantity 1 g
Formula Weight 202.26
Percent Purity 97%
Chemical Name or Material 4-(3-Piperidinyloxy)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'