missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Dimethylamino)propoxy]-N-methylbenzylamine, 90%, Thermo Scientific™

54.70 € - 132.00 €

Chemical Identifiers

CAS 910037-04-6
Molecular Formula C13H22N2O
Molecular Weight (g/mol) 222.332
MDL Number MFCD09064994
InChI Key LFLFEMOINFHORX-UHFFFAOYSA-N
Synonym 4-3-dimethylamino propoxy-n-methylbenzylamine, dimethyl 3-4-methylamino methyl phenoxy propyl amine, n,n-dimethyl-3-4-methylamino methyl phenoxy propan-1-amine, 4-3-dimethylamino propoxy phenyl methyl methyl amine, benzenemethanamine, 4-3-dimethylamino propoxy-n-methyl
PubChem CID 24229610
IUPAC Name N,N-dimethyl-3-[4-(methylaminomethyl)phenoxy]propan-1-amine
SMILES CNCC1=CC=C(C=C1)OCCCN(C)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10632665
View Documents
Thermo Scientific Maybridge
CC59546CB
250 mg Amber Glass Bottle
54.70 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10642665
View Documents
Thermo Scientific Maybridge
CC59546DA
1 g Amber Glass Bottle
132.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

910037-04-6
222.332
LFLFEMOINFHORX-UHFFFAOYSA-N
24229610
CNCC1=CC=C(C=C1)OCCCN(C)C
C13H22N2O
MFCD09064994
4-3-dimethylamino propoxy-n-methylbenzylamine, dimethyl 3-4-methylamino methyl phenoxy propyl amine, n,n-dimethyl-3-4-methylamino methyl phenoxy propan-1-amine, 4-3-dimethylamino propoxy phenyl methyl methyl amine, benzenemethanamine, 4-3-dimethylamino propoxy-n-methyl
N,N-dimethyl-3-[4-(methylaminomethyl)phenoxy]propan-1-amine

Specifications

910037-04-6
100.0
C13H22N2O
4-3-dimethylamino propoxy-n-methylbenzylamine, dimethyl 3-4-methylamino methyl phenoxy propyl amine, n,n-dimethyl-3-4-methylamino methyl phenoxy propan-1-amine, 4-3-dimethylamino propoxy phenyl methyl methyl amine, benzenemethanamine, 4-3-dimethylamino propoxy-n-methyl
CNCC1=CC=C(C=C1)OCCCN(C)C
222.332
222.33
4-[3-(Dimethylamino)propoxy]-N-methylbenzylamine
89.5
90%
MFCD09064994
LFLFEMOINFHORX-UHFFFAOYSA-N
N,N-dimethyl-3-[4-(methylaminomethyl)phenoxy]propan-1-amine
24229610
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.