missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(1-Imidazolyl)propylaminomethyl]benzeneboronic acid pinacol ester, Thermo Scientific™

77.95 € - 277.00 €

Chemical Identifiers

CAS 1257648-78-4
Molecular Formula C19H28BN3O2
Molecular Weight (g/mol) 341.26
MDL Number MFCD12546046
InChI Key OJNUJXDRCCIJCB-UHFFFAOYSA-N
Synonym 4-3-1-imidazolyl propylaminomethyl benzeneboronic acid pinacol ester, 3-imidazol-1-yl-propyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl amine, amtb157, 4-3-1-imidazolyl propylaminomethyl phenylboronic acid pinacol ester, n-3-1-imidazolyl propyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylamine, 3-imidazol-1-yl propyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl amine, 3-imidazol-1-yl propyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl amine
PubChem CID 57416871
IUPAC Name 3-imidazol-1-yl-N-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]propan-1-amine
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(CNCCCN2C=CN=C2)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15429245
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H51671.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15439245
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H51671.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1257648-78-4
341.26
OJNUJXDRCCIJCB-UHFFFAOYSA-N
57416871
CC1(C)OB(OC1(C)C)C1=CC=C(CNCCCN2C=CN=C2)C=C1
C19H28BN3O2
MFCD12546046
4-3-1-imidazolyl propylaminomethyl benzeneboronic acid pinacol ester, 3-imidazol-1-yl-propyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl amine, amtb157, 4-3-1-imidazolyl propylaminomethyl phenylboronic acid pinacol ester, n-3-1-imidazolyl propyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylamine, 3-imidazol-1-yl propyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl amine, 3-imidazol-1-yl propyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl amine
3-imidazol-1-yl-N-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]propan-1-amine

Specifications

1257648-78-4
MFCD12546046
OJNUJXDRCCIJCB-UHFFFAOYSA-N
3-imidazol-1-yl-N-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]propan-1-amine
57416871
4-[3-(1-Imidazolyl)propylaminomethyl]benzeneboronic acid pinacol ester
C19H28BN3O2
4-3-1-imidazolyl propylaminomethyl benzeneboronic acid pinacol ester, 3-imidazol-1-yl-propyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl amine, amtb157, 4-3-1-imidazolyl propylaminomethyl phenylboronic acid pinacol ester, n-3-1-imidazolyl propyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylamine, 3-imidazol-1-yl propyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl amine, 3-imidazol-1-yl propyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl amine
CC1(C)OB(OC1(C)C)C1=CC=C(CNCCCN2C=CN=C2)C=C1
341.26
341.26
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications