missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methoxyethyl)phenol, 98%, Thermo Scientific Chemicals

Product Code. 11485567
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56718-71-9
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00017537
InChI Key FAYGEALAEQKPDI-UHFFFAOYSA-N
Synonym 4-2-methoxyethyl phenol, p-2-methoxyethyl phenol, phenol, 4-2-methoxyethyl, 4-methoxyethylphenol, unii-py1w0akw8i, metoprolol impurity b, metoprolol impurity i, py1w0akw8i, 4-hydroxyphenylethyl methyl ether
PubChem CID 92516
IUPAC Name 4-(2-methoxyethyl)phenol
SMILES COCCC1=CC=C(C=C1)O

Specifications

Melting Point 43°C to 45°C
Quantity 250 g
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 4-(2-Methoxyethyl)phenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.