missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Hydroxyphenyl)-2-(1-piperidinyl)thiazole, 97%, Thermo Scientific™

Chemical Identifiers

CAS 916497-18-2
Molecular Formula C14H16N2OS
Molecular Weight (g/mol) 260.36
MDL Number MFCD22682977
InChI Key CIZFLLFRDXZCLM-UHFFFAOYSA-N
Synonym 2-2-piperidin-1-yl-1,3-thiazol-4-yl phenol, 4-2-hydroxyphenyl-2-1-piperidinyl thiazole
PubChem CID 15984509
IUPAC Name 2-(2-piperidin-1-yl-1,3-thiazol-4-yl)phenol
SMILES OC1=CC=CC=C1C1=CSC(=N1)N1CCCCC1
View More Specs

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

916497-18-2
260.36
CIZFLLFRDXZCLM-UHFFFAOYSA-N
15984509
OC1=CC=CC=C1C1=CSC(=N1)N1CCCCC1
C14H16N2OS
MFCD22682977
2-2-piperidin-1-yl-1,3-thiazol-4-yl phenol, 4-2-hydroxyphenyl-2-1-piperidinyl thiazole
2-(2-piperidin-1-yl-1,3-thiazol-4-yl)phenol

Specifications

916497-18-2
MFCD22682977
CIZFLLFRDXZCLM-UHFFFAOYSA-N
2-(2-piperidin-1-yl-1,3-thiazol-4-yl)phenol
15984509
97%
C14H16N2OS
2-2-piperidin-1-yl-1,3-thiazol-4-yl phenol, 4-2-hydroxyphenyl-2-1-piperidinyl thiazole
OC1=CC=CC=C1C1=CSC(=N1)N1CCCCC1
260.36
289.29
4-(2-Hydroxyphenyl)-2-(1-piperidinyl)thiazole
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications