missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1H-Tetrazol-5-yl)phenol, 97%, Thermo Scientific™
61.31 € - 494.00 €
Chemical Identifiers
CAS | 51517-88-5 |
---|---|
Molecular Formula | C7H6N4O |
Molecular Weight (g/mol) | 162.15 |
MDL Number | MFCD05662709 |
InChI Key | GDDQLGTXWIFSDI-UHFFFAOYSA-N |
Synonym | 4-1h-tetrazol-5-yl phenol, 5-4-hydroxyphenyl tetrazole, 5-4-hydroxyphenyl-1h-tetrazole, 4-1h-1,2,3,4-tetrazol-5-yl phenol, phenol, 4-1h-tetrazol-5-yl, 4-2h-tetrazol-5-yl phenol, 4-2h-tetrazol-5-yl-phenol, qoymnywfhzfxhr-uhfffaoysa-n, 4-2h-1,2,3,4-tetrazol-5-yl phenol, 4-1,2-dihydrotetrazol-5-ylidene cyclohexa-2,5-dien-1-one |
PubChem CID | 5413297 |
SMILES | O=C1C=CC(C=C1)=C1NNN=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15483557
|
Thermo Scientific Alfa Aesar
H26918.MD |
250 mg |
61.31 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11411490
|
Thermo Scientific Alfa Aesar
H26918.03 |
1 g | N/A | ||||||
11421490
|
Thermo Scientific Alfa Aesar
H26918.06 |
5 g | N/A | ||||||
Description
Chemical Identifiers
51517-88-5 | |
162.15 | |
GDDQLGTXWIFSDI-UHFFFAOYSA-N | |
5413297 |
C7H6N4O | |
MFCD05662709 | |
4-1h-tetrazol-5-yl phenol, 5-4-hydroxyphenyl tetrazole, 5-4-hydroxyphenyl-1h-tetrazole, 4-1h-1,2,3,4-tetrazol-5-yl phenol, phenol, 4-1h-tetrazol-5-yl, 4-2h-tetrazol-5-yl phenol, 4-2h-tetrazol-5-yl-phenol, qoymnywfhzfxhr-uhfffaoysa-n, 4-2h-1,2,3,4-tetrazol-5-yl phenol, 4-1,2-dihydrotetrazol-5-ylidene cyclohexa-2,5-dien-1-one | |
O=C1C=CC(C=C1)=C1NNN=N1 |
Specifications
51517-88-5 | |
C7H6N4O | |
4-1h-tetrazol-5-yl phenol, 5-4-hydroxyphenyl tetrazole, 5-4-hydroxyphenyl-1h-tetrazole, 4-1h-1,2,3,4-tetrazol-5-yl phenol, phenol, 4-1h-tetrazol-5-yl, 4-2h-tetrazol-5-yl phenol, 4-2h-tetrazol-5-yl-phenol, qoymnywfhzfxhr-uhfffaoysa-n, 4-2h-1,2,3,4-tetrazol-5-yl phenol, 4-1,2-dihydrotetrazol-5-ylidene cyclohexa-2,5-dien-1-one | |
O=C1C=CC(C=C1)=C1NNN=N1 | |
162.15 | |
162.16 | |
4-(1H-Tetrazol-5-yl)phenol |
∽240°C (decomposition) | |
MFCD05662709 | |
GDDQLGTXWIFSDI-UHFFFAOYSA-N | |
4-(1,2-dihydrotetrazol-5-ylidene)cyclohexa-2,5-dien-1-one | |
5413297 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title