missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-1,2,4-triazol-1-yl)aniline, 97%, Thermo Scientific™

30.60 € - 206.00 €

Chemical Identifiers

CAS 6523-49-5
Molecular Formula C8H8N4
Molecular Weight (g/mol) 160.18
MDL Number MFCD00171470
InChI Key BLNPEJXSNSBBNM-UHFFFAOYSA-N
Synonym 4-1,2,4 triazol-1-yl-phenylamine, 4-1h-1,2,4-triazol-1-yl aniline, 4-1,2,4-triazol-1-yl aniline, 1-4-aminophenyl-1,2,4-triazole, triazolylaniline, 1-4'-aminophenyl-1,2,4-triazole, benzenamine, 4-1h-1,2,4-triazol-1-yl, 4-1h-1,2,4-triazol-1-yl benzenamine, 4-1,2,4-triazolyl phenylamine, pubchem3927
PubChem CID 735826
IUPAC Name 4-(1,2,4-triazol-1-yl)aniline
SMILES C1=CC(=CC=C1N)N2C=NC=N2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10724193
View Documents Promotion Details
Thermo Scientific Maybridge
CC18614DA
1 g
30.60 €
1g
Please to purchase this item. Need a web account? Register with us today!
10049091
View Documents Promotion Details
Thermo Scientific Maybridge
CC18614EA
10 g
206.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

6523-49-5
160.18
BLNPEJXSNSBBNM-UHFFFAOYSA-N
735826
C1=CC(=CC=C1N)N2C=NC=N2
C8H8N4
MFCD00171470
4-1,2,4 triazol-1-yl-phenylamine, 4-1h-1,2,4-triazol-1-yl aniline, 4-1,2,4-triazol-1-yl aniline, 1-4-aminophenyl-1,2,4-triazole, triazolylaniline, 1-4'-aminophenyl-1,2,4-triazole, benzenamine, 4-1h-1,2,4-triazol-1-yl, 4-1h-1,2,4-triazol-1-yl benzenamine, 4-1,2,4-triazolyl phenylamine, pubchem3927
4-(1,2,4-triazol-1-yl)aniline

Specifications

6523-49-5
100.0
Amber Glass Bottle
MFCD00171470
BLNPEJXSNSBBNM-UHFFFAOYSA-N
4-(1,2,4-triazol-1-yl)aniline
735826
97%
96.5
97%
C8H8N4
4-1,2,4 triazol-1-yl-phenylamine, 4-1h-1,2,4-triazol-1-yl aniline, 4-1,2,4-triazol-1-yl aniline, 1-4-aminophenyl-1,2,4-triazole, triazolylaniline, 1-4'-aminophenyl-1,2,4-triazole, benzenamine, 4-1h-1,2,4-triazol-1-yl, 4-1h-1,2,4-triazol-1-yl benzenamine, 4-1,2,4-triazolyl phenylamine, pubchem3927
C1=CC(=CC=C1N)N2C=NC=N2
160.18
160.18
4-(1H-1, 2, 4-Triazol-1-yl)aniline
Videos
SDS
Documents

Documents

Product Certifications