Learn More
3M™ Disposable Protective Overshoe Covers 442
Click to view available options
White
Description
A reliable, disposable boot cover designed to help protect footwear against light liquid splashes and hazardous dusts. Made from a microporous laminate material with an anti-static treatment.
- Covers are elasticated to help ensure a good fit
- Antistatic treatment
- Universal size
Specifications
Specifications
Size | Universal |
Material | Microporous Fabric |
Coating | Microporous Film Laminate |
Color | White |
Disposable | Single-use |
Closure Type | Elastic Ankle |
Model | 442 |
Resistance | Dry Particulates, Fluid |
Product Type | Boot Covers |