missing translation for 'onlineSavingsMsg'
Learn More

3M™ Disposable Protective Coveralls 4540+, Hooded

Product Code. 11903794
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Coveralls are designed to help protect against hazardous dusts (Type 5) and certain light liquid splashes (Type 6).

  • Premium dual material technology provides both protective performance and comfort
  • Barrier against dry particles and limited liquid chemical splash (CE Type 5/6)
  • Soft and light, super low-linting laminated microporous material
  • Back panel made of breathable SMS material for improved air circulation and reduced heat stress
  • Two-way zipper with fully sealable storm flap
  • Three-panel hood design for enhanced compatability with complimentary PPE
  • Knitted cuffs and elasticated waist and ankles for increased comfort and freedom of movement
  • Anti-static treated
  • Low linting
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material Polyethylene, Polyester/Polyethylene Laminate
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit
Neck Style Hooded
Hood Type 3-panel
Closure Type Front Zipper with Storm Flap
Seam Type Serged, Bound
Sterility Non-sterile
Resistance Anti-static, Dust, Liquid
Coating Anti-static
Back Style Breathable SMMMS material
Certifications/Compliance ANSI 101-1996
Certifications/Compliance (Europe) CE Category III Type 5/6, EN 13034, EN-ISO 13982-1, EN 1149-1:2006, EN 1073
Description White Polypropylene/Polyethylene laminated suit, SMMMS Polypropylene blue back panel, Nylon on Polyester braid zipper, Neoprene Rubber elastic, Polyester cuffs and thread
Latex Free No
Model 4540+
Size (English) Chest 39 to 43 in.
Size (English) Inseam 32 in.
Size (English) Waist 51 in.
Size (Metric) Chest 99.06 to 109.22 cm
Size (Metric) Inseam 81.28 cm
Size (Metric) Waist 129.54 cm
For Use With (Application) Pesticide, Asbestos and Lead Abatement, Power Plants, Metals, Cleaning, Maintenance, General Industrial, Insulation, Food Processing, Pharmaceutical
Height (English) 5.75 to 6 ft.
Height (Metric) 174 to 181 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.