missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ White Protective Coverall 4520
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
- Designed to offer protection against hazardous dusts and limited liquid splashes and sprays
- Made from lightweight and breathable material for reduced heat stress and premium wearer comfort all day
- Standard collar for use with respiratory equipment or where a hood may not be desired
- Knitted cuffs and elasticated waist and ankles for increased comfort and freedom of movement
Specifications
Specifications
Product Type | Protective Coveralls |
Size | Small |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Waist Style | Elastic |
Cuff Style | Knit |
Neck Style | Hooded |
Hood Type | 3-panel |
Closure Type | 2-way Zipper with Storm Flap |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'