missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3H,5H-Octamethyltetrasiloxane, 96%, Thermo Scientific Chemicals
48.30 € - 162.00 €
Chemical Identifiers
CAS | 16066-09-4 |
---|---|
Molecular Formula | C8H24O3Si4 |
Molecular Weight (g/mol) | 280.617 |
MDL Number | MFCD00053664 |
InChI Key | OHSYWAVRSCQMHG-UHFFFAOYSA-N |
Synonym | 1,1,1,3,5,7,7,7-octamethyltetrasiloxane, 3h,5h-octamethyltetrasiloxane, 1,3-bis trimethylsiloxy-1,3-dimethyldisiloxane, tetrasiloxane, 1,1,1,3,5,7,7,7-octamethyl, bis trimethylsiloxy dimethyldisiloxane, methyl-methyl trimethylsilyloxy, 1,1,1,3,5,7,7,7-octamethyl-tetrasiloxane, tetrasiloxane, 1,1,1,3,5,7,7,7-octamethyl-,, 1,1,1,3,5,7,7,7-octamethyltetrasiloxane, technical gc |
PubChem CID | 6327318 |
IUPAC Name | methyl-[methyl(trimethylsilyloxy)-$l^{3}-silanyl]oxy-trimethylsilyloxysilicon |
SMILES | C[Si](O[Si](C)O[Si](C)(C)C)O[Si](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11333856
|
Thermo Scientific Alfa Aesar
L16956.06 |
5 g |
48.30 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343856
|
Thermo Scientific Alfa Aesar
L16956.14 |
25 g |
162.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16066-09-4 | |
280.617 | |
OHSYWAVRSCQMHG-UHFFFAOYSA-N | |
6327318 | |
C[Si](O[Si](C)O[Si](C)(C)C)O[Si](C)(C)C |
C8H24O3Si4 | |
MFCD00053664 | |
1,1,1,3,5,7,7,7-octamethyltetrasiloxane, 3h,5h-octamethyltetrasiloxane, 1,3-bis trimethylsiloxy-1,3-dimethyldisiloxane, tetrasiloxane, 1,1,1,3,5,7,7,7-octamethyl, bis trimethylsiloxy dimethyldisiloxane, methyl-methyl trimethylsilyloxy, 1,1,1,3,5,7,7,7-octamethyl-tetrasiloxane, tetrasiloxane, 1,1,1,3,5,7,7,7-octamethyl-,, 1,1,1,3,5,7,7,7-octamethyltetrasiloxane, technical gc | |
methyl-[methyl(trimethylsilyloxy)-$l^{3}-silanyl]oxy-trimethylsilyloxysilicon |
Specifications
169°C | |
C8H24O3Si4 | |
78°C (172°F) | |
OHSYWAVRSCQMHG-UHFFFAOYSA-N | |
methyl-[methyl(trimethylsilyloxy)-$l^{3}-silanyl]oxy-trimethylsilyloxysilicon | |
6327318 | |
96% | |
Moisture sensitive | |
3H,5H-Octamethyltetrasiloxane |
16066-09-4 | |
MFCD00053664 | |
1,1,1,3,5,7,7,7-octamethyltetrasiloxane, 3h,5h-octamethyltetrasiloxane, 1,3-bis trimethylsiloxy-1,3-dimethyldisiloxane, tetrasiloxane, 1,1,1,3,5,7,7,7-octamethyl, bis trimethylsiloxy dimethyldisiloxane, methyl-methyl trimethylsilyloxy, 1,1,1,3,5,7,7,7-octamethyl-tetrasiloxane, tetrasiloxane, 1,1,1,3,5,7,7,7-octamethyl-,, 1,1,1,3,5,7,7,7-octamethyltetrasiloxane, technical gc | |
C[Si](O[Si](C)O[Si](C)(C)C)O[Si](C)(C)C | |
280.617 | |
282.64 | |
1.386 | |
0.863 |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
missing translation for 'einecsNumber' : 240-209-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only