Learn More
Sampling Systems™ 316 Stainless Steel Micro Spatula
Sampling Systems™ 316 Stainless Steel Micro Spatula is ideal for sampling or dispensing small volumes of powder.
Brand:  Sampling Systems™ A106-20
Product Code. 15839581
  • 41.75 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless steel
  • For powder sampling
  • Crevice free
  • Bright polish finish
Specifications
Specifications
Micro Spatula
12 mm
175 mm
40 mm
Stainless Steel