missing translation for 'onlineSavingsMsg'
Learn More

(±)-3-(Trifluoromethyl)piperidine, 98%, Thermo Scientific™

Product Code. 15214408
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 768-31-0
Molecular Formula C6H11F3N
Molecular Weight (g/mol) 154.16
MDL Number MFCD02183561
InChI Key JOHFJTBDUSVGQB-RXMQYKEDSA-O
Synonym 3-trifluoromethyl piperidine, dl-3-trifluoromethyl piperidine, piperidine, 3-trifluoromethyl, 3-trifluoromethyl-piperidine, 3-trifluormethyl-piperidine, piperidine,3-trifluoromethyl
PubChem CID 2760776
IUPAC Name 3-(trifluoromethyl)piperidine
SMILES FC(F)(F)[C@@H]1CCC[NH2+]C1

Specifications

Boiling Point 128.0°C to 130.0°C
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 153.15
Percent Purity 98%
Chemical Name or Material (±)-3-(Trifluoromethyl)piperidine
compliance-icons compliance-icons
Product Identifier
  • (±)-3-(Trifluoromethyl)piperidine, 98%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'