Learn More
3-Trifluoromethoxyphenylboronic acid, 98%, Thermo Scientific™
38.80 € - 108.00 €
Chemical Identifiers
CAS | 179113-90-7 |
---|---|
Molecular Formula | C7H6BF3O3 |
Molecular Weight (g/mol) | 205.93 |
MDL Number | MFCD01320697 |
InChI Key | UWDFWVLAHRQSKK-UHFFFAOYSA-N |
Synonym | 3-trifluoromethoxyphenylboronic acid, 3-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boronic acid, 3-trifluoromethoxy benzeneboronic acid, m-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boranediol, 3-trifluoromethyloxy phenylboronic acid, boronic acid, 3-trifluoromethoxy phenyl |
PubChem CID | 2734385 |
IUPAC Name | [3-(trifluoromethoxy)phenyl]boronic acid |
SMILES | OB(O)C1=CC=CC(OC(F)(F)F)=C1 |
Description
Chemical Identifiers
179113-90-7 | |
205.93 | |
UWDFWVLAHRQSKK-UHFFFAOYSA-N | |
2734385 | |
OB(O)C1=CC=CC(OC(F)(F)F)=C1 |
C7H6BF3O3 | |
MFCD01320697 | |
3-trifluoromethoxyphenylboronic acid, 3-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boronic acid, 3-trifluoromethoxy benzeneboronic acid, m-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boranediol, 3-trifluoromethyloxy phenylboronic acid, boronic acid, 3-trifluoromethoxy phenyl | |
[3-(trifluoromethoxy)phenyl]boronic acid |
Specifications
179113-90-7 | |
100.0 | |
Beige to White | |
97.5% min. (HPLC) | |
C7H6BF3O3 | |
3-trifluoromethoxyphenylboronic acid, 3-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boronic acid, 3-trifluoromethoxy benzeneboronic acid, m-trifluoromethoxy phenylboronic acid, 3-trifluoromethoxy phenyl boranediol, 3-trifluoromethyloxy phenylboronic acid, boronic acid, 3-trifluoromethoxy phenyl | |
OB(O)C1=CC=CC(OC(F)(F)F)=C1 | |
205.93 | |
205.93 | |
Chunks and/or Powder |
97.5 | |
79°C | |
Authentic | |
Glass Bottle | |
MFCD01320697 | |
UWDFWVLAHRQSKK-UHFFFAOYSA-N | |
[3-(trifluoromethoxy)phenyl]boronic acid | |
2734385 | |
98% | |
3-Trifluoromethoxyphenylboronic acid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- 3-Trifluoromethoxyphenylboronic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.