missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Trifluoromethoxy)benzoyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-(Trifluoromethoxy)benzoyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Incompatible with oxidizing agents, bases and alcohols.
3-(Trifluoromethoxy)benzoyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Incompatible with oxidizing agents, bases and alcohols.
Specifications
Specifications
Refractive Index | 1.467 |
Quantity | 1 g |
UN Number | UN3265 |
Beilstein | 7006050 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 224.57 |
Percent Purity | 97% |
Chemical Name or Material | 3-(Trifluoromethoxy)benzoyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'