missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ 3-Stop Tubing, Tygon LMT-55
Designed for optimal operation in your Ismatec pump.
Brand: Masterflex™ SC0065T
This item is not returnable.
View return policy
Description
- For use with pumps that incorporate cassettes into the pumphead design.
- Color-coded stops.
- Inexpensive all-around tubing for general laboratory applications.
- Transparent.
- Resistant to almost all inorganic chemicals.
- Smooth polished inner wall.
- Low gas permeability.
- Non-aging and non-oxidizing.
Specifications
Yellow-Blue | |
Tygon LMT-55 | |
General laboratory applications, pumps with cassettes in pumphead design | |
12/Pk | |
I.D. 1.52 mm | |
74°C |
1.52 mm | |
400 mm | |
−50°C | |
Acids: Good, Alkaline solutions: Good, Solvents: Not rec., Pressure: Fair, Vacuum: Good, Viscous media: Excellent, Sterile media: Limited | |
Transparent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction