missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Piperidinobenzaldehyde, 95%, Thermo Scientific™
51.50 € - 278.00 €
Chemical Identifiers
CAS | 669050-72-0 |
---|---|
Molecular Formula | C12H15NO |
Molecular Weight (g/mol) | 189.258 |
InChI Key | FXXQUTSXXILOMY-UHFFFAOYSA-N |
Synonym | 3-piperidinobenzaldehyde, 3-piperidin-1-yl benzaldehyde, benzaldehyde, 3-1-piperidinyl, 3-piperidylbenzaldehyde, 3-1-piperidinyl benzaldehyde |
PubChem CID | 7164587 |
IUPAC Name | 3-piperidin-1-ylbenzaldehyde |
SMILES | C1CCN(CC1)C2=CC=CC(=C2)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10452083
|
Thermo Scientific Maybridge
CC34004CB |
250 mg | Amber Glass Bottle |
51.50 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10525963
|
Thermo Scientific Maybridge
CC34004DA |
1 g | Amber Glass Bottle |
98.20 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10133724
|
Thermo Scientific Maybridge
CC34004DE |
5 g | Amber Glass Bottle |
278.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
669050-72-0 | |
189.258 | |
3-piperidinobenzaldehyde, 3-piperidin-1-yl benzaldehyde, benzaldehyde, 3-1-piperidinyl, 3-piperidylbenzaldehyde, 3-1-piperidinyl benzaldehyde | |
3-piperidin-1-ylbenzaldehyde |
C12H15NO | |
FXXQUTSXXILOMY-UHFFFAOYSA-N | |
7164587 | |
C1CCN(CC1)C2=CC=CC(=C2)C=O |
Specifications
669050-72-0 | |
C12H15NO | |
FXXQUTSXXILOMY-UHFFFAOYSA-N | |
3-piperidin-1-ylbenzaldehyde | |
7164587 | |
95% |
95% | |
3-piperidinobenzaldehyde, 3-piperidin-1-yl benzaldehyde, benzaldehyde, 3-1-piperidinyl, 3-piperidylbenzaldehyde, 3-1-piperidinyl benzaldehyde | |
C1CCN(CC1)C2=CC=CC(=C2)C=O | |
189.258 | |
189.26 | |
3-Piperidinobenzaldehyde |