missing translation for 'onlineSavingsMsg'
Learn More

3-(Phenylamino)propyltrimethoxysilane, 96%, Thermo Scientific Chemicals

29.25 € - 69.90 €

Chemical Identifiers

CAS 3068-76-6
Molecular Formula C12H21NO3Si
Molecular Weight (g/mol) 255.389
MDL Number MFCD00053673
InChI Key KBJFYLLAMSZSOG-UHFFFAOYSA-N
Synonym n-3-trimethoxysilyl propyl aniline, trimethoxy 3-phenylamino propyl silane, n-phenylaminopropyltrimethoxysilane, unii-7au1h1fv6a, benzenamine, n-3-trimethoxysilyl propyl, n-phenyl-3-aminopropyltrimethoxysilane, 7au1h1fv6a, n-3-trimethoxysilylpropyl aniline, 3-phenylamino propyltrimethoxysilane
PubChem CID 76476
IUPAC Name N-(3-trimethoxysilylpropyl)aniline
SMILES CO[Si](CCCNC1=CC=CC=C1)(OC)OC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11343796
View Documents
Thermo Scientific Alfa Aesar
L16850.14
25 g
29.25 €
25g
Please to purchase this item. Need a web account? Register with us today!
11353796
View Documents
Thermo Scientific Alfa Aesar
L16850.22
100 g
69.90 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3068-76-6
255.389
KBJFYLLAMSZSOG-UHFFFAOYSA-N
76476
CO[Si](CCCNC1=CC=CC=C1)(OC)OC
C12H21NO3Si
MFCD00053673
n-3-trimethoxysilyl propyl aniline, trimethoxy 3-phenylamino propyl silane, n-phenylaminopropyltrimethoxysilane, unii-7au1h1fv6a, benzenamine, n-3-trimethoxysilyl propyl, n-phenyl-3-aminopropyltrimethoxysilane, 7au1h1fv6a, n-3-trimethoxysilylpropyl aniline, 3-phenylamino propyltrimethoxysilane
N-(3-trimethoxysilylpropyl)aniline

Specifications

310°C
C12H21NO3Si
MFCD00053673
8620045
n-3-trimethoxysilyl propyl aniline, trimethoxy 3-phenylamino propyl silane, n-phenylaminopropyltrimethoxysilane, unii-7au1h1fv6a, benzenamine, n-3-trimethoxysilyl propyl, n-phenyl-3-aminopropyltrimethoxysilane, 7au1h1fv6a, n-3-trimethoxysilylpropyl aniline, 3-phenylamino propyltrimethoxysilane
CO[Si](CCCNC1=CC=CC=C1)(OC)OC
255.389
255.39
Weak
Moisture sensitive
3-(Phenylamino)propyltrimethoxysilane
3068-76-6
C6H5NH(CH2)3Si(OCH3)3
UN3267
165°C (329°F)
KBJFYLLAMSZSOG-UHFFFAOYSA-N
N-(3-trimethoxysilylpropyl)aniline
76476
96%
1.5055
1.07
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H351-H373
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 221-328-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only