Learn More
3-Oxocyclobutanecarboxylic acid, 98%, Thermo Scientific Chemicals
73.97 € - 295.47 €
Chemical Identifiers
CAS | 23761-23-1 |
---|---|
Molecular Formula | C5H6O3 |
Molecular Weight (g/mol) | 114.10 |
MDL Number | MFCD00100900 |
InChI Key | IENOFRJPUPTEMI-UHFFFAOYSA-N |
Synonym | 3-oxocyclobutanecarboxylic acid, 3-oxo-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 3-oxo, 3-oxo-cyclobutanecarboxylicacid, 1-carboxy-3-oxocyclobutane, 3-oxocyclobutyl carboxylic acid, 3-oxocyclobutanecarboxylicacid, pubchem14220, acmc-1cak7, ksc201q6h |
PubChem CID | 4913358 |
IUPAC Name | 3-oxocyclobutane-1-carboxylic acid |
SMILES | OC(=O)C1CC(=O)C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15472119
|
Thermo Scientific Alfa Aesar
H64774.06 |
5 g |
73.97 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15482119
|
Thermo Scientific Alfa Aesar
H64774.14 |
25 g |
295.47 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Useful intermediate for pharmaceutical, organic synthesis and organic light-emitting diode(OLED).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUseful intermediate for pharmaceutical, organic synthesis and organic light-emitting diode(OLED).
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
23761-23-1 | |
114.10 | |
IENOFRJPUPTEMI-UHFFFAOYSA-N | |
4913358 | |
OC(=O)C1CC(=O)C1 |
C5H6O3 | |
MFCD00100900 | |
3-oxocyclobutanecarboxylic acid, 3-oxo-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 3-oxo, 3-oxo-cyclobutanecarboxylicacid, 1-carboxy-3-oxocyclobutane, 3-oxocyclobutyl carboxylic acid, 3-oxocyclobutanecarboxylicacid, pubchem14220, acmc-1cak7, ksc201q6h | |
3-oxocyclobutane-1-carboxylic acid |
Specifications
23761-23-1 | |
White | |
MFCD00100900 | |
3-oxocyclobutanecarboxylic acid, 3-oxo-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 3-oxo, 3-oxo-cyclobutanecarboxylicacid, 1-carboxy-3-oxocyclobutane, 3-oxocyclobutyl carboxylic acid, 3-oxocyclobutanecarboxylicacid, pubchem14220, acmc-1cak7, ksc201q6h | |
IENOFRJPUPTEMI-UHFFFAOYSA-N | |
3-oxocyclobutane-1-carboxylic acid | |
4913358 | |
98% | |
3-Oxocyclobutanecarboxylic acid |
69°C to 70°C | |
C5H6O3 | |
2078401 | |
Slightly soluble in water. | |
OC(=O)C1CC(=O)C1 | |
114.10 | |
114.1 | |
Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.