Learn More
3-Nitrophthalic anhydride, 97%, Thermo Scientific Chemicals
47.84 € - 133.67 €
Chemical Identifiers
CAS | 641-70-3 |
---|---|
Molecular Formula | C8H3NO5 |
Molecular Weight (g/mol) | 193.11 |
MDL Number | MFCD00005921 |
InChI Key | ROFZMKDROVBLNY-UHFFFAOYSA-N |
Synonym | 3-nitrophthalic anhydride, 4-nitroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-nitro, phthalic anhydride, 3-nitro, 3-nitro phthalic anhydride, 3-nitrophthalic acid anhydride, 1,3-isobenzofurandione, nitro, 4-nitro-isobenzofuran-1,3-dione, 3-nitrophthalicanhydride, nitrophthalic anhydride |
PubChem CID | 21631 |
IUPAC Name | 4-nitro-2-benzofuran-1,3-dione |
SMILES | [O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10645032
|
Thermo Scientific Acros
168990250 |
25 g | Glass bottle |
47.84 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10285690
|
Thermo Scientific Acros
168991000 |
100 g | Glass bottle |
133.67 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
641-70-3 | |
193.11 | |
ROFZMKDROVBLNY-UHFFFAOYSA-N | |
21631 | |
[O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O |
C8H3NO5 | |
MFCD00005921 | |
3-nitrophthalic anhydride, 4-nitroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-nitro, phthalic anhydride, 3-nitro, 3-nitro phthalic anhydride, 3-nitrophthalic acid anhydride, 1,3-isobenzofurandione, nitro, 4-nitro-isobenzofuran-1,3-dione, 3-nitrophthalicanhydride, nitrophthalic anhydride | |
4-nitro-2-benzofuran-1,3-dione |
Specifications
641-70-3 | |
100.0 | |
Beige to Yellow | |
97% | |
MFCD00005921 | |
3-nitrophthalic anhydride, 4-nitroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-nitro, phthalic anhydride, 3-nitro, 3-nitro phthalic anhydride, 3-nitrophthalic acid anhydride, 1,3-isobenzofurandione, nitro, 4-nitro-isobenzofuran-1,3-dione, 3-nitrophthalicanhydride, nitrophthalic anhydride | |
ROFZMKDROVBLNY-UHFFFAOYSA-N | |
4-nitro-2-benzofuran-1,3-dione | |
21631 | |
97% | |
3-Nitrophthalic anhydride, 97% |
96.0 | |
160.0°C to 165.0°C | |
Authentic | |
C8H3NO5 | |
17, 486 | |
Solubility in water: decomposes. | |
[O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O | |
193.11 | |
193.12 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 211-373-6
missing translation for 'tsca' : TSCA
- 3-Nitrophthalic anhydride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.