Learn More
3-Nitrophenylboronic acid, 97%, Thermo Scientific Chemicals
30.20 € - 84.50 €
Chemical Identifiers
CAS | 13331-27-6 |
---|---|
Molecular Formula | C6H6BNO4 |
Molecular Weight (g/mol) | 166.93 |
MDL Number | MFCD00007193 |
InChI Key | ZNRGSYUVFVNSAW-UHFFFAOYSA-N |
Synonym | 3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol |
PubChem CID | 1677 |
IUPAC Name | (3-nitrophenyl)boronic acid |
SMILES | OB(O)C1=CC=CC(=C1)[N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10215491
|
Thermo Scientific Acros
344700010 |
1 g | Glass bottle |
30.20 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10419871
|
Thermo Scientific Acros
344700050 |
5 g | Glass bottle |
84.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13331-27-6 | |
166.93 | |
ZNRGSYUVFVNSAW-UHFFFAOYSA-N | |
1677 | |
OB(O)C1=CC=CC(=C1)[N+]([O-])=O |
C6H6BNO4 | |
MFCD00007193 | |
3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol | |
(3-nitrophenyl)boronic acid |
Spezifikation
13331-27-6 | |
100.0 | |
Brown to Yellow | |
97% | |
NO2C6H4B(OH)2 | |
3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol | |
ZNRGSYUVFVNSAW-UHFFFAOYSA-N | |
(3-nitrophenyl)boronic acid | |
1677 | |
97% | |
3-Nitrophenylboronic acid, 97% |
96.0 | |
289.0°C to 291.0°C | |
Authentic | |
C6H6BNO4 | |
MFCD00007193 | |
Solubility in water: soluble in hot water. Other solubilities: soluble in methanol | |
OB(O)C1=CC=CC(=C1)[N+]([O-])=O | |
166.93 | |
166.93 | |
Powder |
Sicherheit und Handhabung
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning
missing translation for 'rtecsNumber' : CY8980000
missing translation for 'tsca' : TSCA
- 3-Nitrophenylboronic acid
Signalwort
- Warning
Gefahrenkategorie
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Gefahrenhinweis
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Sicherheitshinweis
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only