missing translation for 'onlineSavingsMsg'
Learn More

3-(n-Butylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15459437
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 397843-70-8
Molecular Formula C11H16BNO3
Molecular Weight (g/mol) 221.063
MDL Number MFCD04115691
InChI Key UMTGJEHDHSYNMS-UHFFFAOYSA-N
Synonym 3-butylcarbamoyl phenyl boronic acid, 3-butylaminocarbonyl phenylboronic acid, 3-butylcarbamoyl phenylboronic acid, 3-butylaminocarbonyl benzeneboronic acid, 3-n-butylcarbamoyl benzeneboronic acid, 3-butylaminocarbonyl phenylboronicacid, 3-n-butylaminocarbonyl phenylboronic acid, boronic acid, 3-butylamino carbonyl phenyl, acmc-209j7o
PubChem CID 3693263
IUPAC Name [3-(butylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)C(=O)NCCCC)(O)O

Specifications

Melting Point 216°C to 224°C
Quantity 250 mg
Formula Weight 221.06
Percent Purity 97%
Chemical Name or Material 3-(n-Butylcarbamoyl)benzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.