missing translation for 'onlineSavingsMsg'
Learn More

3-n-Butoxypropionitrile, 97%, Thermo Scientific™

28.00 € - 81.00 €

Chemical Identifiers

CAS 6959-71-3
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.19
MDL Number MFCD00045643
InChI Key AWVNJBFNHGQUQU-UHFFFAOYSA-N
Synonym 3-butoxypropionitrile, propanenitrile, 3-butoxy, beta-butoxy propionitrile, propionitrile, 3-butoxy, 3-butoxypropannitril, 3-butoxypropiononitrile, 3-n-butoxypropionitrile, 3-butoxypropannitril czech, .beta.-butoxy propionitrile, 3-butoxypropanitrile
PubChem CID 23395
IUPAC Name 3-butoxypropanenitrile
SMILES CCCCOCCC#N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11397716
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L11342.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11307726
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L11342.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6959-71-3
127.19
AWVNJBFNHGQUQU-UHFFFAOYSA-N
23395
CCCCOCCC#N
C7H13NO
MFCD00045643
3-butoxypropionitrile, propanenitrile, 3-butoxy, beta-butoxy propionitrile, propionitrile, 3-butoxy, 3-butoxypropannitril, 3-butoxypropiononitrile, 3-n-butoxypropionitrile, 3-butoxypropannitril czech, .beta.-butoxy propionitrile, 3-butoxypropanitrile
3-butoxypropanenitrile

Specifications

6959-71-3
105°C to 106°C (15 mmHg)
C7H13NO
MFCD00045643
3-butoxypropionitrile, propanenitrile, 3-butoxy, beta-butoxy propionitrile, propionitrile, 3-butoxy, 3-butoxypropannitril, 3-butoxypropiononitrile, 3-n-butoxypropionitrile, 3-butoxypropannitril czech, .beta.-butoxy propionitrile, 3-butoxypropanitrile
CCCCOCCC#N
127.19
127.19
3-n-Butoxypropionitrile
0.92
89°C (192°F)
1.4175
1747904
AWVNJBFNHGQUQU-UHFFFAOYSA-N
3-butoxypropanenitrile
23395
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

missing translation for 'einecsNumber' : 230-153-0

missing translation for 'rtecsNumber' : UG1225000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications