missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiophene, 98+%, Thermo Scientific Chemicals

Product Code. 11425403
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-44-4
Molecular Formula C5H6S
Molecular Weight (g/mol) 98.163
MDL Number MFCD00005470
InChI Key QENGPZGAWFQWCZ-UHFFFAOYSA-N
Synonym thiophene, 3-methyl, 3-thiotolene, beta-methylthiophene, poly, p3mt, 3-methyl thiophene, 3-methyl-thiophene, methyl-3-thiophene, ccris 2937, thiophene, 3-methyl-, homopolymer
PubChem CID 12024
IUPAC Name 3-methylthiophene
SMILES CC1=CSC=C1

Specifications

Melting Point -69°C
Density 1.02
Boiling Point 115°C to 117°C
Flash Point 11°C (51°F)
Odor Aromatic
Refractive Index 1.52
Quantity 500 g
UN Number UN1993
Beilstein 1300
Formula Weight 98.17
Percent Purity ≥98%
Chemical Name or Material 3-Methylthiophene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.