Learn More
3-Methylisoxazole-5-carboxylic acid, 97%, Thermo Scientific™
51.50 € - 119.00 €
Chemical Identifiers
CAS | 4857-42-5 |
---|---|
Molecular Formula | C5H5NO3 |
Molecular Weight (g/mol) | 127.099 |
MDL Number | MFCD00464222 |
InChI Key | HXIYCKAAQPHZBM-UHFFFAOYSA-N |
Synonym | 3-methylisoxazole-5-carboxylic acid, 3-methyl-5-isoxazolecarboxylic acid, 5-isoxazolecarboxylic acid, 3-methyl, 3-methyl-isoxazole-5-carboxylic acid, 5-carboxy-3-methylisoxazole, 5-isoxazolecarboxylicacid, 3-methyl, 3-methylisoxazole-5-carboxylicacid, acmc-1aruu, 3-methyl-5-carboxyisoxazole, aronis24519 |
PubChem CID | 853085 |
IUPAC Name | 3-methyl-1,2-oxazole-5-carboxylic acid |
SMILES | CC1=NOC(=C1)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10432494
|
Thermo Scientific Maybridge
CC27001CB |
250 mg | Amber Glass Bottle |
51.50 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10474815
|
Thermo Scientific Maybridge
CC27001DA |
1 g | Amber Glass Bottle |
119.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
4857-42-5 | |
127.099 | |
HXIYCKAAQPHZBM-UHFFFAOYSA-N | |
853085 | |
CC1=NOC(=C1)C(=O)O |
C5H5NO3 | |
MFCD00464222 | |
3-methylisoxazole-5-carboxylic acid, 3-methyl-5-isoxazolecarboxylic acid, 5-isoxazolecarboxylic acid, 3-methyl, 3-methyl-isoxazole-5-carboxylic acid, 5-carboxy-3-methylisoxazole, 5-isoxazolecarboxylicacid, 3-methyl, 3-methylisoxazole-5-carboxylicacid, acmc-1aruu, 3-methyl-5-carboxyisoxazole, aronis24519 | |
3-methyl-1,2-oxazole-5-carboxylic acid |
Specifications
4857-42-5 | |
100.0 | |
C5H5NO3 | |
3-methylisoxazole-5-carboxylic acid, 3-methyl-5-isoxazolecarboxylic acid, 5-isoxazolecarboxylic acid, 3-methyl, 3-methyl-isoxazole-5-carboxylic acid, 5-carboxy-3-methylisoxazole, 5-isoxazolecarboxylicacid, 3-methyl, 3-methylisoxazole-5-carboxylicacid, acmc-1aruu, 3-methyl-5-carboxyisoxazole, aronis24519 | |
CC1=NOC(=C1)C(=O)O | |
127.099 | |
127.1 | |
3-Methylisoxazole-5-carboxylic acid |
96.5 | |
97% | |
MFCD00464222 | |
HXIYCKAAQPHZBM-UHFFFAOYSA-N | |
3-methyl-1,2-oxazole-5-carboxylic acid | |
853085 | |
97% |
Safety and Handling
GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.