missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15419907
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 832695-88-2
Molecular Formula C8H10BNO3
Molecular Weight (g/mol) 178.98
MDL Number MFCD04038918
InChI Key FYFFPNFUVMBPRZ-UHFFFAOYSA-N
Synonym 3-n-methylaminocarbonyl phenylboronic acid, 3-methylcarbamoyl phenyl boronic acid, 3-methylcarbamoyl benzeneboronic acid, 3-methylcarbamoyl phenylboronic acid, 3-n-methylaminocarbonyl benzeneboronic acid, 3-borono-n-methylbenzamide, boronic acid, 3-methylamino carbonyl phenyl, acmc-209prq, ksc496e8l
PubChem CID 2773524
IUPAC Name [3-(methylcarbamoyl)phenyl]boronic acid
SMILES CNC(=O)C1=CC=CC(=C1)B(O)O

Specifications

Melting Point 114°C to 124°C
Quantity 1 g
Formula Weight 178.98
Percent Purity 98%
Chemical Name or Material 3-(Methylcarbamoyl)benzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'