missing translation for 'onlineSavingsMsg'
Learn More

(3-Methylaminopropyl)trimethoxysilane, 95%, Thermo Scientific Chemicals

Product Code. 11389225
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3069-25-8
Molecular Formula C7H19NO3Si
Molecular Weight (g/mol) 193.32
MDL Number MFCD00048021
InChI Key DVYVMJLSUSGYMH-UHFFFAOYSA-N
Synonym n-methylaminopropyltrimethoxysilane, n-methyl-3-trimethoxysilyl propan-1-amine, trimethoxy 3-methylamino propyl silane, 1-propanamine, n-methyl-3-trimethoxysilyl, n-methyl-3-trimethoxysilyl propylamine, n-methyl-3-aminopropyltrimethoxysilane, 3-methylaminopropyl-trimethoxysilane, methyl 3-trimethoxysilyl propyl amine, n-methyl-3-aminopropyl-trimethoxysilane, acmc-20ak27
PubChem CID 76480
IUPAC Name N-methyl-3-trimethoxysilylpropan-1-amine
SMILES CNCCC[Si](OC)(OC)OC

Specifications

Boiling Point 106°C (30mmHg)
Quantity 100 g
Beilstein 2072720
Flash Point 82°C (179°F)
Solubility Information Hydrolyzes in water.
Formula Weight 193.32
Percent Purity 95%
Refractive Index 1.419
Sensitivity Moisture sensitive
Density 0.978
Chemical Name or Material (3-Methylaminopropyl)trimethoxysilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.