Learn More
3-Methyl-2-cyclopenten-1-one, 98%, stab. with 0.1% hydroquinone, Thermo Scientific Chemicals
81.20 € - 283.41 €
Chemical Identifiers
CAS | 2758-18-1 |
---|---|
Molecular Formula | C6H8O |
Molecular Weight (g/mol) | 96.129 |
MDL Number | MFCD00001403 |
InChI Key | CHCCBPDEADMNCI-UHFFFAOYSA-N |
Synonym | 3-methyl-2-cyclopenten-1-one, 3-methylcyclopent-2-enone, 2-cyclopenten-1-one, 3-methyl, 3-methyl-2-cyclopentenone, 1-methyl-1-cyclopenten-3-one, unii-6v7rsw7273, fema no. 3435, 3-methylcyclopentenone, acmc-1crym, 3-methyl-2cyclopenten-1-one |
PubChem CID | 17691 |
IUPAC Name | 3-methylcyclopent-2-en-1-one |
SMILES | CC1=CC(=O)CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11429070
|
Thermo Scientific Alfa Aesar
A13993.09 |
10 g |
81.20 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439070
|
Thermo Scientific Alfa Aesar
A13993.18 |
50 g |
283.41 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
Solubility
Fully miscible in water.
Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, heat. Store in refrigerator.
Chemical Identifiers
2758-18-1 | |
96.129 | |
CHCCBPDEADMNCI-UHFFFAOYSA-N | |
17691 | |
CC1=CC(=O)CC1 |
C6H8O | |
MFCD00001403 | |
3-methyl-2-cyclopenten-1-one, 3-methylcyclopent-2-enone, 2-cyclopenten-1-one, 3-methyl, 3-methyl-2-cyclopentenone, 1-methyl-1-cyclopenten-3-one, unii-6v7rsw7273, fema no. 3435, 3-methylcyclopentenone, acmc-1crym, 3-methyl-2cyclopenten-1-one | |
3-methylcyclopent-2-en-1-one |
Specifications
2758-18-1 | |
0.98 | |
72°C to 74°C (15 mmHg) | |
Pleasant | |
1.489 | |
1280476 | |
Fully miscible in water. | |
CC1=CC(=O)CC1 | |
96.129 | |
96.13 | |
3-Methyl-2-cyclopenten-1-one, stabilized with 0.1% hydroquinone |
2°C | |
4.9 | |
65°C (149°F) | |
C6H8O | |
MFCD00001403 | |
3-methyl-2-cyclopenten-1-one, 3-methylcyclopent-2-enone, 2-cyclopenten-1-one, 3-methyl, 3-methyl-2-cyclopentenone, 1-methyl-1-cyclopenten-3-one, unii-6v7rsw7273, fema no. 3435, 3-methylcyclopentenone, acmc-1crym, 3-methyl-2cyclopenten-1-one | |
CHCCBPDEADMNCI-UHFFFAOYSA-N | |
3-methylcyclopent-2-en-1-one | |
17691 | |
98% |
Safety and Handling
P210-P235-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H227-H319-H335
missing translation for 'einecsNumber' : 220-421-5
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.