Learn More
(+/-)-3-Methyl-2-butanol, 98%, Thermo Scientific Chemicals
28.34 € - 70.95 €
Chemical Identifiers
CAS | 598-75-4 |
---|---|
Molecular Formula | C5H12O |
Molecular Weight (g/mol) | 88.15 |
MDL Number | MFCD00004527 |
InChI Key | MXLMTQWGSQIYOW-UHFFFAOYNA-N |
Synonym | 3-methyl-2-butanol, sec-isoamyl alcohol, 2-butanol, 3-methyl, 2-methyl-3-butanol, isopropylmethylcarbinol, methylisopropylcarbinol, 1,2-dimethylpropanol, 1,2-dimethyl-1-propanol, dl-3-methyl-2-butanol, 3-methyl-butan-2-ol |
PubChem CID | 11732 |
ChEBI | CHEBI:77517 |
IUPAC Name | 3-methylbutan-2-ol |
SMILES | CC(C)C(C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11377405
|
Thermo Scientific Alfa Aesar
L05347.14 |
25 g |
28.34 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387405
|
Thermo Scientific Alfa Aesar
L05347.22 |
100 g |
70.95 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Methyl-2-butanol has been used as a reference for microbially produced, volatile organic compounds (MVOC) in GC-MS analyses. It acts as a plant metabolite and polar solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methyl-2-butanol has been used as a reference for microbially produced, volatile organic compounds (MVOC) in GC-MS analyses. It acts as a plant metabolite and polar solvent.
Solubility
Miscible in Ether, Alcohols. Slightly soluble in water. (2.8g/100mL, 30°C)
Notes
Incompatible materials are Strong oxidizing agents, Reducing agents.
Chemical Identifiers
598-75-4 | |
88.15 | |
MXLMTQWGSQIYOW-UHFFFAOYNA-N | |
11732 | |
3-methylbutan-2-ol |
C5H12O | |
MFCD00004527 | |
3-methyl-2-butanol, sec-isoamyl alcohol, 2-butanol, 3-methyl, 2-methyl-3-butanol, isopropylmethylcarbinol, methylisopropylcarbinol, 1,2-dimethylpropanol, 1,2-dimethyl-1-propanol, dl-3-methyl-2-butanol, 3-methyl-butan-2-ol | |
CHEBI:77517 | |
CC(C)C(C)O |
Specifications
598-75-4 | |
26°C (78°F) | |
MFCD00004527 | |
1718800 | |
3-methyl-2-butanol, sec-isoamyl alcohol, 2-butanol, 3-methyl, 2-methyl-3-butanol, isopropylmethylcarbinol, methylisopropylcarbinol, 1,2-dimethylpropanol, 1,2-dimethyl-1-propanol, dl-3-methyl-2-butanol, 3-methyl-butan-2-ol | |
CC(C)C(C)O | |
88.15 | |
CHEBI:77517 | |
98% |
112°C | |
C5H12O | |
UN1105 | |
14,6031 | |
MXLMTQWGSQIYOW-UHFFFAOYNA-N | |
3-methylbutan-2-ol | |
11732 | |
88.15 | |
(+/-)-3-Methyl-2-butanol |
Safety and Handling
GHS H Statement
H226-H332-H335
Flammable liquid and vapor.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS
missing translation for 'tsca' : TSCA
RUO – Research Use Only