Learn More
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%, Thermo Scientific Chemicals
46.13 € - 417.08 €
Chemical Identifiers
CAS | 149022-15-1 |
---|---|
Molecular Formula | C8H10ClN3S |
Molecular Weight (g/mol) | 215.70 |
MDL Number | MFCD00149370 |
InChI Key | OEZPVSPULCMUQB-UHFFFAOYSA-N |
Synonym | unii-9u2c4w7epr, mbth hydrochloride, 9u2c4w7epr, 3-methyl-2-benzothiazolone hydrazone hydrochloride, 3-methyl-2-benzothiazolinone hydrazone hydrochloride, z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride, mbth hcl, ccris 1543, 2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1, 2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride |
PubChem CID | 9575839 |
IUPAC Name | (Z)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrochloride |
SMILES | Cl.CN1C(SC2=CC=CC=C12)=NN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11437787
|
Thermo Scientific Alfa Aesar
A16317.06 |
5 g |
46.13 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11447787
|
Thermo Scientific Alfa Aesar
A16317.14 |
25 g |
162.81 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11457787
|
Thermo Scientific Alfa Aesar
A16317.22 |
100 g |
417.08 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate is used in the synthesis of benzothiazolium azo dyes. It is a chromogenic reagent used for the determination of cholesterol, benzodiazepines and enzyme activity. As an electrophilic coupling reagent, it is used in the spectrophotometric determination of chloramine-B (CAB) and other residual chlorine content in various environmental water samples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate is used in the synthesis of benzothiazolium azo dyes. It is a chromogenic reagent used for the determination of cholesterol, benzodiazepines and enzyme activity. As an electrophilic coupling reagent, it is used in the spectrophotometric determination of chloramine-B (CAB) and other residual chlorine content in various environmental water samples.
Solubility
Soluble in water, dimethyl sulfoxide and methanol.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
149022-15-1 | |
215.70 | |
OEZPVSPULCMUQB-UHFFFAOYSA-N | |
9575839 | |
Cl.CN1C(SC2=CC=CC=C12)=NN |
C8H10ClN3S | |
MFCD00149370 | |
unii-9u2c4w7epr, mbth hydrochloride, 9u2c4w7epr, 3-methyl-2-benzothiazolone hydrazone hydrochloride, 3-methyl-2-benzothiazolinone hydrazone hydrochloride, z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride, mbth hcl, ccris 1543, 2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1, 2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride | |
(Z)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrochloride |
Specifications
149022-15-1 | |
C8H10ClN3S | |
UN2811 | |
unii-9u2c4w7epr, mbth hydrochloride, 9u2c4w7epr, 3-methyl-2-benzothiazolone hydrazone hydrochloride, 3-methyl-2-benzothiazolinone hydrazone hydrochloride, z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride, mbth hcl, ccris 1543, 2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1, 2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride | |
OEZPVSPULCMUQB-UHFFFAOYSA-N | |
(Z)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrochloride | |
9575839 | |
97% |
∼275°C (decomposition) | |
MFCD00149370 | |
3571041 | |
Soluble in water,dimethyl sulfoxide and methanol. | |
Cl.CN1C(SC2=CC=CC=C12)=NN | |
215.70 | |
215.71 (Anhydrous) | |
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 238-428-7
missing translation for 'rtecsNumber' : DL7160200
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- 3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
- MIXTURE LIST-Contains : 3-Methyl-2-benzothiazolinone hydrazone hydrochloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.