missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1,2-cyclopentanediol, mixture of diastereomers, 95%, Thermo Scientific™

Product Code. 11484397
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27583-37-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00019285
InChI Key KANFKJUPLALTDB-UHFFFAOYNA-N
Synonym 3-methyl-1,2-cyclopentanediol, 1,2-cyclopentanediol, 3-methyl, 3-methylcyclopentan-1,2-diol, 3-methyl-1,2-cyclopentanediol, mixture of diastereomers, 3-methylcyclopentanediol, acmc-20an1b, 3-methyl-1,2-cyclopentanediol #, 3-methyl-1,2-cyclopentanediol mixture of isomers
PubChem CID 33945
IUPAC Name 3-methylcyclopentane-1,2-diol
SMILES CC1CCC(O)C1O

Specifications

Quantity 25 g
Formula Weight 116.16
Percent Purity 95%
Chemical Name or Material 3-Methyl-1,2-cyclopentanediol, mixture of diastereomers
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.