missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenol, 97%, Thermo Scientific Chemicals

25.93 € - 232.16 €

Chemical Identifiers

CAS 150-19-6
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00002267
InChI Key ASHGTJPOSUFTGB-UHFFFAOYSA-N
Synonym m-methoxyphenol, 3-hydroxyanisole, m-hydroxyanisole, phenol, 3-methoxy, m-guaiacol, resorcinol monomethyl ether, resorcinol methyl ether, 1-hydroxy-3-methoxybenzene, phenol, m-methoxy, 3-methoxy phenol
PubChem CID 9007
ChEBI CHEBI:52678
IUPAC Name 3-methoxyphenol
SMILES COC1=CC=CC(=C1)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11455213
View Documents
Thermo Scientific Alfa Aesar
A12826.14
25 g
25.93 €
25g
Please to purchase this item. Need a web account? Register with us today!
11465213
View Documents
Thermo Scientific Alfa Aesar
A12826.22
100 g
66.93 €
100g
Please to purchase this item. Need a web account? Register with us today!
11475213
View Documents
Thermo Scientific Alfa Aesar
A12826.36
500 g
232.16 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in the synthesis of C(4) symmetric calix[4]resorcinarene, 2-nitroso-5-methoxyphenol and 6-methoxy-2(3H)-benzoxazolone. It is also used as a flavoring agent. They are also used are used to make antioxidants and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of C(4) symmetric calix[4]resorcinarene, 2-nitroso-5-methoxyphenol and 6-methoxy-2(3H)-benzoxazolone. It is also used as a flavoring agent. They are also used are used to make antioxidants and pharmaceuticals.

Solubility
Partly soluble in water.

Notes
Store away from oxidizing agents, acids, bases, acid chlorides and acid anhydrides. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources
Specifications

Chemical Identifiers

150-19-6
124.139
ASHGTJPOSUFTGB-UHFFFAOYSA-N
9007
3-methoxyphenol
C7H8O2
MFCD00002267
m-methoxyphenol, 3-hydroxyanisole, m-hydroxyanisole, phenol, 3-methoxy, m-guaiacol, resorcinol monomethyl ether, resorcinol methyl ether, 1-hydroxy-3-methoxybenzene, phenol, m-methoxy, 3-methoxy phenol
CHEBI:52678
COC1=CC=CC(=C1)O

Specifications

150-19-6
1.14
124°C (255°F)
C7H8O2
MFCD00002267
1209898
Partly soluble in water.
COC1=CC=CC(=C1)O
124.139
CHEBI:52678
97%
-18°C
244°C
Sweet
1.552
UN2810
m-methoxyphenol, 3-hydroxyanisole, m-hydroxyanisole, phenol, 3-methoxy, m-guaiacol, resorcinol monomethyl ether, resorcinol methyl ether, 1-hydroxy-3-methoxybenzene, phenol, m-methoxy, 3-methoxy phenol
ASHGTJPOSUFTGB-UHFFFAOYSA-N
3-methoxyphenol
9007
124.14
3-Methoxyphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H318-H302-H332-H315-H335
Toxic in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c

H302+H332-H311-H315-H318-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 205-754-6

missing translation for 'rtecsNumber' : SL7524000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • 3-Methoxyphenol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : 3-Methoxyphenol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.