missing translation for 'onlineSavingsMsg'
Learn More

3'-Methoxyacetophenone, 97%, Thermo Scientific Chemicals

Product Code. 11449160
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is applied in the asymmetric transfer hydrogenation of ketones catalyzed by ruthenium(II) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in the asymmetric transfer hydrogenation of ketones catalyzed by ruthenium(II) complexes.

Solubility
Fully miscible in water. Soluble in alcohol.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-37-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00008736
InChI Key BAYUSCHCCGXLAY-UHFFFAOYSA-N
Synonym 3'-methoxyacetophenone, 3-methoxyacetophenone, 3-acetylanisole, m-methoxyacetophenone, 1-3-methoxyphenyl ethanone, ethanone, 1-3-methoxyphenyl, m-acetanisole, acetophenone, m-methoxy, 1-3-methoxyphenyl ethan-1-one, acetophenone, 3'-methoxy
PubChem CID 11460
IUPAC Name 1-(3-methoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)OC

Specifications

Melting Point -7°C
Density 1.094
Boiling Point 239°C to 241°C
Flash Point >110°C (230°F)
Refractive Index 1.5425
Quantity 100 g
Beilstein 878552
Sensitivity Light sensitive
Solubility Information Fully miscible in water. Soluble in alcohol.
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material 3'-Methoxyacetophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'