Learn More
(3-Mercaptopropyl)triethoxysilane, 94%, Thermo Scientific Chemicals
81.90 € - 256.00 €
Chemical Identifiers
CAS | 14814-09-6 |
---|---|
Molecular Formula | C9H22O3SSi |
Molecular Weight (g/mol) | 238.417 |
MDL Number | MFCD00053924 |
InChI Key | DCQBZYNUSLHVJC-UHFFFAOYSA-N |
Synonym | 3-mercaptopropyltriethoxysilane, 3-mercaptopropyl triethoxysilane, 3-triethoxysilyl propane-1-thiol, 1-propanethiol, 3-triethoxysilyl, 3-triethoxysilyl propanethiol, unii-z069rx894n, triethoxy 3-mercaptopropyl silane, 3-triethoxysilyl propyl mercaptan, gamma-mercaptopropyl triethoxysilane, silane, 3-mercaptopropyl triethoxy |
PubChem CID | 84658 |
IUPAC Name | 3-triethoxysilylpropane-1-thiol |
SMILES | CCO[Si](CCCS)(OCC)OCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11494687
|
Thermo Scientific Alfa Aesar
B21191.14 |
25 g |
81.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404697
|
Thermo Scientific Alfa Aesar
B21191.22 |
100 g |
256.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It can improve the properties of the mineral-filled elastomer, including modulus, tensile and tear strength, heat buildup, abrasion resistance, resilience. It can be used as a pretreatment on minerals or added at the time of compounding. Mineral-reinforced rubbers such as shoe soles, rollers, wheels, white sidewalls, wire and cable insulation are the common case (3-Mercaptopropyl)triethoxysilane, could be used in. It also finds it uses as an intermediate, in polymer and tire manufacture, and as a laboratory reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt can improve the properties of the mineral-filled elastomer, including modulus, tensile and tear strength, heat buildup, abrasion resistance, resilience. It can be used as a pretreatment on minerals or added at the time of compounding. Mineral-reinforced rubbers such as shoe soles, rollers, wheels, white sidewalls, wire and cable insulation are the common case (3-Mercaptopropyl)triethoxysilane, could be used in. It also finds it uses as an intermediate, in polymer and tire manufacture, and as a laboratory reagent.
Solubility
Hydrolyzes slowly in water.
Notes
Air & moisture sensitive. Store away from oxidizing agents, air and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
14814-09-6 | |
238.417 | |
DCQBZYNUSLHVJC-UHFFFAOYSA-N | |
84658 | |
CCO[Si](CCCS)(OCC)OCC |
C9H22O3SSi | |
MFCD00053924 | |
3-mercaptopropyltriethoxysilane, 3-mercaptopropyl triethoxysilane, 3-triethoxysilyl propane-1-thiol, 1-propanethiol, 3-triethoxysilyl, 3-triethoxysilyl propanethiol, unii-z069rx894n, triethoxy 3-mercaptopropyl silane, 3-triethoxysilyl propyl mercaptan, gamma-mercaptopropyl triethoxysilane, silane, 3-mercaptopropyl triethoxy | |
3-triethoxysilylpropane-1-thiol |
Specifications
210°C | |
C9H22O3SSi | |
MFCD00053924 | |
88°C (190°F) | |
Hydrolyzes slowly in water. | |
CCO[Si](CCCS)(OCC)OCC | |
238.417 | |
238.42 | |
Unpleasant | |
Air and moisture sensitive | |
(3-Mercaptopropyl)triethoxysilane |
14814-09-6 | |
(CH3CH2O)3Si(CH2)3SH | |
2039575 | |
3-mercaptopropyltriethoxysilane, 3-mercaptopropyl triethoxysilane, 3-triethoxysilyl propane-1-thiol, 1-propanethiol, 3-triethoxysilyl, 3-triethoxysilyl propanethiol, unii-z069rx894n, triethoxy 3-mercaptopropyl silane, 3-triethoxysilyl propyl mercaptan, gamma-mercaptopropyl triethoxysilane, silane, 3-mercaptopropyl triethoxy | |
DCQBZYNUSLHVJC-UHFFFAOYSA-N | |
3-triethoxysilylpropane-1-thiol | |
84658 | |
94% | |
1.437 | |
0.98 |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 238-883-1
missing translation for 'rtecsNumber' : TZ7760000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only