Learn More
(3-Mercaptopropyl)methyldimethoxysilane, 95%, Thermo Scientific Chemicals
20.20 € - 626.00 €
Chemical Identifiers
CAS | 31001-77-1 |
---|---|
Molecular Formula | C6H16O2SSi |
Molecular Weight (g/mol) | 180.337 |
MDL Number | MFCD00053923 |
InChI Key | IKYAJDOSWUATPI-UHFFFAOYSA-N |
Synonym | 3-mercaptopropylmethyldimethoxysilane, 3-mercaptopropylmethyldimethoxy silane, 3-dimethoxymethylsilyl-1-propanethiol, 3-mercaptopropyl methyldimethoxysilane, dimethoxy-3-mercaptopropyl methylsilane, 3-dimethoxy methyl silyl propane-1-thiol, acmc-1aiak, ksc496q0j, 3-methyldimethoxysilyl propylmercaptan |
PubChem CID | 2759524 |
IUPAC Name | 3-[dimethoxy(methyl)silyl]propane-1-thiol |
SMILES | CO[Si](C)(CCCS)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11344598
|
Thermo Scientific Alfa Aesar
042426.AB |
10 mL | |||||||
11324598
|
Thermo Scientific Alfa Aesar
042426.AD |
50 mL |
57.50 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11354598
|
Thermo Scientific Alfa Aesar
042426.AK |
250 mL |
184.00 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11334598
|
Thermo Scientific Alfa Aesar
042426.K2 |
1 L | |||||||
Description
(3-Mercaptopropyl)methyldimethoxysilane is used as coupling agent to improve the adhesion of sulfur cured elastomers (polysulfide, polyurethane sealants) to inorganic fillers (such as silica, glass, clay, mica), fiberglass and surfaces. It is also used as a pretreatment on minerals or added at the time of compounding and to improve low-rolling resistance in silica-reinforced tire tread compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
31001-77-1 | |
180.337 | |
IKYAJDOSWUATPI-UHFFFAOYSA-N | |
2759524 | |
CO[Si](C)(CCCS)OC |
C6H16O2SSi | |
MFCD00053923 | |
3-mercaptopropylmethyldimethoxysilane, 3-mercaptopropylmethyldimethoxy silane, 3-dimethoxymethylsilyl-1-propanethiol, 3-mercaptopropyl methyldimethoxysilane, dimethoxy-3-mercaptopropyl methylsilane, 3-dimethoxy methyl silyl propane-1-thiol, acmc-1aiak, ksc496q0j, 3-methyldimethoxysilyl propylmercaptan | |
3-[dimethoxy(methyl)silyl]propane-1-thiol |
Specifications
96°C (30mmHg) | |
Colorless | |
C6H16O2SSi | |
MFCD00053923 | |
3-mercaptopropylmethyldimethoxysilane, 3-mercaptopropylmethyldimethoxy silane, 3-dimethoxymethylsilyl-1-propanethiol, 3-mercaptopropyl methyldimethoxysilane, dimethoxy-3-mercaptopropyl methylsilane, 3-dimethoxy methyl silyl propane-1-thiol, acmc-1aiak, ksc496q0j, 3-methyldimethoxysilyl propylmercaptan | |
IKYAJDOSWUATPI-UHFFFAOYSA-N | |
3-[dimethoxy(methyl)silyl]propane-1-thiol | |
2759524 | |
95% | |
Moisture sensitive | |
(3-Mercaptopropyl)methyldimethoxysilane |
31001-77-1 | |
Liquid | |
HSCH2CH2CH2Si(OCH3)2CH3 | |
93°C (199°F) | |
Insoluble in water. | |
CO[Si](C)(CCCS)OC | |
180.337 | |
180.34 | |
1.45 | |
1.000 g/mL |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P330-P333+P313-P363-P370+P378q-P501c
H227-H302-H317
missing translation for 'einecsNumber' : 250-426-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only