missing translation for 'onlineSavingsMsg'
Learn More

3-Iodopropionic acid, 99%, Thermo Scientific Chemicals

24.92 € - 78.69 €

Chemical Identifiers

CAS 141-76-4
Molecular Formula C3H5IO2
Molecular Weight (g/mol) 199.98
MDL Number MFCD00002765
InChI Key KMRNTNDWADEIIX-UHFFFAOYSA-N
Synonym 3-iodopropionic acid, propanoic acid, 3-iodo, beta-iodopropionic acid, propionic acid, 3-iodo, iodopropanoic acid, 3-iodo-propionic acid, .beta.-iodopropionic acid, iodo-propanoic acid, b-iodopropionic acid, 3-iodo-propanoic acid
PubChem CID 8856
IUPAC Name 3-iodopropanoic acid
SMILES OC(=O)CCI
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11327785
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L02655.06
5 g
24.92 €
5g
Please to purchase this item. Need a web account? Register with us today!
11337785
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L02655.14
25 g
78.69 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

141-76-4
199.98
KMRNTNDWADEIIX-UHFFFAOYSA-N
8856
OC(=O)CCI
C3H5IO2
MFCD00002765
3-iodopropionic acid, propanoic acid, 3-iodo, beta-iodopropionic acid, propionic acid, 3-iodo, iodopropanoic acid, 3-iodo-propionic acid, .beta.-iodopropionic acid, iodo-propanoic acid, b-iodopropionic acid, 3-iodo-propanoic acid
3-iodopropanoic acid

Specifications

141-76-4
C3H5IO2
UN3261
Light sensitive
KMRNTNDWADEIIX-UHFFFAOYSA-N
3-iodopropanoic acid
8856
99%
80°C to 83°C
MFCD00002765
1699514
3-iodopropionic acid, propanoic acid, 3-iodo, beta-iodopropionic acid, propionic acid, 3-iodo, iodopropanoic acid, 3-iodo-propionic acid, .beta.-iodopropionic acid, iodo-propanoic acid, b-iodopropionic acid, 3-iodo-propanoic acid
OC(=O)CCI
199.98
199.98
3-Iodopropionic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 205-499-0

missing translation for 'rtecsNumber' : UF4900000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only