missing translation for 'onlineSavingsMsg'
Learn More

3-Indoxyl sulfate potassium salt, 97%, Thermo Scientific Chemicals

Product Code. 11488946
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2642-37-7
Molecular Formula C8H6KNO4S
Molecular Weight (g/mol) 251.297
MDL Number MFCD00037931
InChI Key MDAWATNFDJIBBD-UHFFFAOYSA-M
Synonym potassium 1h-indol-3-yl sulfate, urinary indican, potassium indol-3-yl sulfate, indican urinary, indol-3-yl potassium sulfate, indoxyl sulfate potassium salt, potassium indol-3-yl sulphate, unii-567hmw942w, indol-3-ol, potassium sulfate, indol-3-yl sulfate, potassium salt
PubChem CID 5177095
IUPAC Name potassium;1H-indol-3-yl sulfate
SMILES C1=CC=C2C(=C1)C(=CN2)OS(=O)(=O)[O-].[K+]

Specifications

Melting Point 165°C (decomposition)
Quantity 1 g
Beilstein 3921324
Sensitivity Light sensitive
Formula Weight 251.31
Percent Purity 97%
Chemical Name or Material 3-Indoxyl sulfate potassium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'