missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals

23.10 € - 228.00 €

Chemical Identifiers

CAS 621-37-4
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00004337
InChI Key FVMDYYGIDFPZAX-UHFFFAOYSA-N
Synonym 3-hydroxyphenylacetic acid, 2-3-hydroxyphenyl acetic acid, 3-hydroxyphenyl acetic acid, 3-hydroxybenzeneacetic acid, 3-hydroxyphenylacetate, m-hydroxyphenylacetic acid, m-hydroxyphenyl acetic acid, benzeneacetic acid, 3-hydroxy, metahydroxy phenylacetic acid, acetic acid, m-hydroxyphenyl
PubChem CID 12122
ChEBI CHEBI:17445
IUPAC Name 2-(3-hydroxyphenyl)acetic acid
SMILES OC(=O)CC1=CC=CC(O)=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11465587
View Documents
Thermo Scientific Alfa Aesar
B23563.03
1 g
23.10 €
1g
Please to purchase this item. Need a web account? Register with us today!
11475587
View Documents
Thermo Scientific Alfa Aesar
B23563.06
5 g
68.40 €
5g
Please to purchase this item. Need a web account? Register with us today!
11485587
View Documents
Thermo Scientific Alfa Aesar
B23563.14
25 g
228.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

621-37-4
152.15
FVMDYYGIDFPZAX-UHFFFAOYSA-N
12122
2-(3-hydroxyphenyl)acetic acid
C8H8O3
MFCD00004337
3-hydroxyphenylacetic acid, 2-3-hydroxyphenyl acetic acid, 3-hydroxyphenyl acetic acid, 3-hydroxybenzeneacetic acid, 3-hydroxyphenylacetate, m-hydroxyphenylacetic acid, m-hydroxyphenyl acetic acid, benzeneacetic acid, 3-hydroxy, metahydroxy phenylacetic acid, acetic acid, m-hydroxyphenyl
CHEBI:17445
OC(=O)CC1=CC=CC(O)=C1

Specifications

621-37-4
C8H8O3
2086506
FVMDYYGIDFPZAX-UHFFFAOYSA-N
2-(3-hydroxyphenyl)acetic acid
12122
152.15
3-Hydroxyphenylacetic acid
128°C to 132°C
MFCD00004337
3-hydroxyphenylacetic acid, 2-3-hydroxyphenyl acetic acid, 3-hydroxyphenyl acetic acid, 3-hydroxybenzeneacetic acid, 3-hydroxyphenylacetate, m-hydroxyphenylacetic acid, m-hydroxyphenyl acetic acid, benzeneacetic acid, 3-hydroxy, metahydroxy phenylacetic acid, acetic acid, m-hydroxyphenyl
OC(=O)CC1=CC=CC(O)=C1
152.15
CHEBI:17445
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 210-684-4

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.