Learn More
3-Hydroxy-2-butanone, monomer + dimer, 95%, Thermo Scientific Chemicals
49.95 € - 460.29 €
Chemical Identifiers
CAS | 513-86-0 |
---|---|
Molecular Formula | C4H8O2 |
Molecular Weight (g/mol) | 88.11 |
MDL Number | MFCD00004521,MFCD00038696 |
InChI Key | ROWKJAVDOGWPAT-UHFFFAOYNA-N |
Synonym | acetoin, 3-hydroxy-2-butanone, acetyl methyl carbinol, dimethylketol, 2-butanone, 3-hydroxy, acetylmethylcarbinol, 2,3-butanolone, 2-hydroxy-3-butanone, 1-hydroxyethyl methyl ketone, methanol, acetylmethyl |
PubChem CID | 179 |
ChEBI | CHEBI:15688 |
IUPAC Name | 3-hydroxybutan-2-one |
SMILES | CC(O)C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11424794
|
Thermo Scientific Alfa Aesar
A13752.18 |
50 g |
49.95 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434794
|
Thermo Scientific Alfa Aesar
A13752.30 |
250 g |
142.71 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414794
|
Thermo Scientific Alfa Aesar
A13752.0B |
1000 g |
460.29 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Hydroxy-2-butanone is a chemical used in food flavoring and fragrances. It acts as an intermediate of butanediol cycle in microorganisms. It is used as an aroma carrier in the preparation of flavors and essences.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Hydroxy-2-butanone is a chemical used in food flavoring and fragrances. It acts as an intermediate of butanediol cycle in microorganisms. It is used as an aroma carrier in the preparation of flavors and essences.
Solubility
Miscible with water, propylene glycol and alcohol. Slightly miscible with ether, petroleum ether. Immiscible with fatty oils and vegetable oil.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
513-86-0 | |
88.11 | |
ROWKJAVDOGWPAT-UHFFFAOYNA-N | |
179 | |
3-hydroxybutan-2-one |
C4H8O2 | |
MFCD00004521,MFCD00038696 | |
acetoin, 3-hydroxy-2-butanone, acetyl methyl carbinol, dimethylketol, 2-butanone, 3-hydroxy, acetylmethylcarbinol, 2,3-butanolone, 2-hydroxy-3-butanone, 1-hydroxyethyl methyl ketone, methanol, acetylmethyl | |
CHEBI:15688 | |
CC(O)C(C)=O |
Specifications
513-86-0 | |
148°C | |
Characteristic | |
MFCD00004521,MFCD00038696 | |
385636 | |
acetoin, 3-hydroxy-2-butanone, acetyl methyl carbinol, dimethylketol, 2-butanone, 3-hydroxy, acetylmethylcarbinol, 2,3-butanolone, 2-hydroxy-3-butanone, 1-hydroxyethyl methyl ketone, methanol, acetylmethyl | |
ROWKJAVDOGWPAT-UHFFFAOYNA-N | |
3-hydroxybutan-2-one | |
179 | |
88.11 | |
3-Hydroxy-2-butanone, monomer + dimer |
1.013 | |
41°C (106°F) | |
C4H8O2 | |
UN2621 | |
14,64 | |
Miscible with water,propylene glycol and alcohol. Slightly miscible with ether,petroleum ether. Immiscible with fatty oils and vegetable oil. | |
CC(O)C(C)=O | |
88.11 | |
CHEBI:15688 | |
95% |
Safety and Handling
GHS H Statement
H226-H315-H319
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
H226-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ACETYL METHYL CARBINOL
missing translation for 'einecsNumber' : 208-174-1
missing translation for 'rtecsNumber' : EL8790000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
- 3-Hydroxy-2-butanone, monomer + dimer
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Acetyl methyl carbinol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.