Learn More
3-Furaldehyde, 98%, stabilized, Thermo Scientific Chemicals
28.14 € - 261.30 €
Chemical Identifiers
CAS | 498-60-2 |
---|---|
Molecular Formula | C5H4O2 |
Molecular Weight (g/mol) | 96.08 |
InChI Key | AZVSIHIBYRHSLB-UHFFFAOYSA-N |
Synonym | 3-furaldehyde, 3-furancarboxaldehyde, furan-3-carboxaldehyde, 3-formylfuran, 3-furfural, unii-pob632x444, 3furaldehyde, 3-furancarbaldehyde, pubchem6948 |
PubChem CID | 10351 |
ChEBI | CHEBI:87609 |
IUPAC Name | furan-3-carbaldehyde |
SMILES | C1=COC=C1C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11327631
|
Thermo Scientific Acros
393260010 |
1 g | Glass bottle |
28.14 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11956171
|
Thermo Scientific Acros
393260050 |
5 g | Glass bottle |
81.71 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11966171
|
Thermo Scientific Acros
393260250 |
25 g | Glass bottle |
261.30 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
498-60-2 | |
96.08 | |
3-furaldehyde, 3-furancarboxaldehyde, furan-3-carboxaldehyde, 3-formylfuran, 3-furfural, unii-pob632x444, 3furaldehyde, 3-furancarbaldehyde, pubchem6948 | |
CHEBI:87609 | |
C1=COC=C1C=O |
C5H4O2 | |
AZVSIHIBYRHSLB-UHFFFAOYSA-N | |
10351 | |
furan-3-carbaldehyde |
Specifications
498-60-2 | |
100.0 | |
1.1500g/mL | |
48°C | |
97.5% min. (GC) | |
1.4930 to 1.4950 (20°C, 589nm) | |
3-furaldehyde, 3-furancarboxaldehyde, furan-3-carboxaldehyde, 3-formylfuran, 3-furfural, unii-pob632x444, 3furaldehyde, 3-furancarbaldehyde, pubchem6948 | |
C1=COC=C1C=O | |
96.08 | |
CHEBI:87609 | |
98% | |
3-Furaldehyde |
97.5 | |
Yellow to Brown | |
145.0°C | |
Authentic | |
C5H4O2 | |
1.15 | |
AZVSIHIBYRHSLB-UHFFFAOYSA-N | |
furan-3-carbaldehyde | |
10351 | |
96.08 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Toxic if swallowed.
Fatal if inhaled.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable f
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.- No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathi
- 3-Furaldehyde
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.