missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15475325
Click to view available options
Quantity:
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 307496-28-2
Density 0.995
Molecular Formula C6H4FIZn
MDL Number MFCD01311395
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym 3-fluorophenyl zincylium iodide, zinc, 3-fluorophenyl iodo, grdtucmqmqtybs-uhfffaoysa-m
InChI Key KXRVGYUTTUSSHQ-UHFFFAOYSA-M
SMILES C1=CC(=C[C-]=C1)F.[Zn+]I
IUPAC Name fluorobenzene;iodozinc(1+)
Molecular Weight (g/mol) 287.381
PubChem CID 6093571
Formula Weight 287.38
Chemical Name or Material 3-Fluorophenylzinc iodide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.