missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-N-(4-fluorobenzyl)aniline, 97%, Thermo Scientific™

Product Code. 15413548
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1019613-18-3
Molecular Formula C13H11F2N
Molecular Weight (g/mol) 219.24
MDL Number MFCD11144475
InChI Key HOWHCAHRSXJDGW-UHFFFAOYSA-N
Synonym 3-fluoro-n-4-fluorobenzyl aniline, 3-fluoro-n-4-fluorophenyl methyl aniline
PubChem CID 28455758
IUPAC Name 3-fluoro-N-[(4-fluorophenyl)methyl]aniline
SMILES FC1=CC=C(CNC2=CC(F)=CC=C2)C=C1

Specifications

Quantity 1 g
Formula Weight 219.23
Percent Purity 97%
Chemical Name or Material 3-Fluoro-N-(4-fluorobenzyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'