Learn More
3-Ethyl-3-oxetanemethanol, 96%, Thermo Scientific™
19.90 € - 163.50 €
Chemical Identifiers
CAS | 3047-32-3 |
---|---|
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00190143 |
InChI Key | UNMJLQGKEDTEKJ-UHFFFAOYSA-N |
Synonym | 3-ethyl-3-oxetanemethanol, 3-ethyloxetan-3-yl methanol, 3-oxetanemethanol, 3-ethyl, 3-ethyloxetane-3-methanol, unii-622325vbyo, 3-ethyl-3-hydroxymethyloxetane, 3-hydroxymethyl-3-ethyloxetane, 3-ethyl-3-hydroxymethyl oxetane, 3-ethyl-oxetan-3-yl-methanol, 2-hydroxymethyl-2-ethyl-1,3-epoxypropane |
PubChem CID | 76444 |
IUPAC Name | (3-ethyloxetan-3-yl)methanol |
SMILES | CCC1(COC1)CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10130613
|
Thermo Scientific Acros
377700010 |
1 g | Glass bottle | N/A | |||||
10108802
|
Thermo Scientific Acros
377700050 |
5 g | Glass bottle | N/A | |||||
10668034
|
Thermo Scientific Acros
377700250 |
25 g | Glass bottle | N/A | |||||
Description
Chemical Identifiers
3047-32-3 | |
MFCD00190143 | |
3-ethyl-3-oxetanemethanol, 3-ethyloxetan-3-yl methanol, 3-oxetanemethanol, 3-ethyl, 3-ethyloxetane-3-methanol, unii-622325vbyo, 3-ethyl-3-hydroxymethyloxetane, 3-hydroxymethyl-3-ethyloxetane, 3-ethyl-3-hydroxymethyl oxetane, 3-ethyl-oxetan-3-yl-methanol, 2-hydroxymethyl-2-ethyl-1,3-epoxypropane | |
(3-ethyloxetan-3-yl)methanol |
116.16 | |
UNMJLQGKEDTEKJ-UHFFFAOYSA-N | |
76444 | |
CCC1(COC1)CO |
Specifications
3047-32-3 | |
100.0 | |
1.0190g/mL | |
108°C | |
95% min. (GC) | |
1.4520 to 1.4540 | |
1.019 | |
UNMJLQGKEDTEKJ-UHFFFAOYSA-N | |
(3-ethyloxetan-3-yl)methanol | |
76444 | |
96% | |
3-Ethyl-3-oxetanemethanol |
95.0 | |
Colorless | |
96.0°C (4.0 mmHg) | |
Authentic | |
C6H12O2 | |
MFCD00190143 | |
3-ethyl-3-oxetanemethanol, 3-ethyloxetan-3-yl methanol, 3-oxetanemethanol, 3-ethyl, 3-ethyloxetane-3-methanol, unii-622325vbyo, 3-ethyl-3-hydroxymethyloxetane, 3-hydroxymethyl-3-ethyloxetane, 3-ethyl-3-hydroxymethyl oxetane, 3-ethyl-oxetan-3-yl-methanol, 2-hydroxymethyl-2-ethyl-1,3-epoxypropane | |
CCC1(COC1)CO | |
116.16 | |
116.16 | |
Liquid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 221-254-
- 3-Ethyl-3-oxetanemethanol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only