missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxyphenol, 98%, Thermo Scientific Chemicals

48.70 € - 527.00 €

Chemical Identifiers

CAS 621-34-1
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00016450
InChI Key VBIKLMJHBGFTPV-UHFFFAOYSA-N
Synonym m-ethoxyphenol, phenol, 3-ethoxy, phenol, m-ethoxy, resorcinol monoethyl ether, 3-ethoxy-phenol, unii-iky4o3474a, 3-ethyloxy phenol, pubchem9510, acmc-209mzu, phenol, m-ethoxy-8ci
PubChem CID 69306
IUPAC Name 3-ethoxyphenol
SMILES CCOC1=CC=CC(=C1)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11421327
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B24266.06
5 g
48.70 €
5g
Please to purchase this item. Need a web account? Register with us today!
11431327
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B24266.14
25 g
151.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
11441327
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B24266.22
100 g
527.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

621-34-1
138.166
VBIKLMJHBGFTPV-UHFFFAOYSA-N
69306
CCOC1=CC=CC(=C1)O
C8H10O2
MFCD00016450
m-ethoxyphenol, phenol, 3-ethoxy, phenol, m-ethoxy, resorcinol monoethyl ether, 3-ethoxy-phenol, unii-iky4o3474a, 3-ethyloxy phenol, pubchem9510, acmc-209mzu, phenol, m-ethoxy-8ci
3-ethoxyphenol

Specifications

621-34-1
1.07
>93°C (199°F)
C8H10O2
MFCD00016450
VBIKLMJHBGFTPV-UHFFFAOYSA-N
3-ethoxyphenol
69306
98%
28°C to 31°C
247°C
Phenol-like
1.541
m-ethoxyphenol, phenol, 3-ethoxy, phenol, m-ethoxy, resorcinol monoethyl ether, 3-ethoxy-phenol, unii-iky4o3474a, 3-ethyloxy phenol, pubchem9510, acmc-209mzu, phenol, m-ethoxy-8ci
CCOC1=CC=CC(=C1)O
138.166
138.17
3-Ethoxyphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

missing translation for 'einecsNumber' : 210-681-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only