Learn More
3-Ethoxy-2-cyclohexen-1-one, 99%, Thermo Scientific Chemicals
47.10 € - 771.00 €
Chemical Identifiers
CAS | 5323-87-5 |
---|---|
Molecular Formula | C8H12O2 |
Molecular Weight (g/mol) | 140.18 |
MDL Number | MFCD00001580 |
InChI Key | JWCFJPLIRVYENQ-UHFFFAOYSA-N |
Synonym | 3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone |
PubChem CID | 79216 |
IUPAC Name | 3-ethoxycyclohex-2-en-1-one |
SMILES | CCOC1=CC(=O)CCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11482327
|
Thermo Scientific Alfa Aesar
A14224.09 |
10 g |
47.10 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11492327
|
Thermo Scientific Alfa Aesar
A14224.18 |
50 g |
186.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11402337
|
Thermo Scientific Alfa Aesar
A14224.30 |
250 g |
771.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Ethoxy-2-cyclohexen-1-one is a useful intermediate for the preparation of 3-alkyl- or aryl-2-cyclohexen-1-ones, by addition of organometallic reagents and subsequent hydrolysis. It helps in kinetic enolate formation (LDA) and alkylation occurs at the 6-position, the Stork-Danheiser kinetic alkylation procedure, which has found extensive use in alicyclic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Ethoxy-2-cyclohexen-1-one is a useful intermediate for the preparation of 3-alkyl- or aryl-2-cyclohexen-1-ones, by addition of organometallic reagents and subsequent hydrolysis. It helps in kinetic enolate formation (LDA) and alkylation occurs at the 6-position, the Stork-Danheiser kinetic alkylation procedure, which has found extensive use in alicyclic synthesis.
Solubility
Slightly soluble in water.
Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
5323-87-5 | |
140.18 | |
JWCFJPLIRVYENQ-UHFFFAOYSA-N | |
79216 | |
CCOC1=CC(=O)CCC1 |
C8H12O2 | |
MFCD00001580 | |
3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone | |
3-ethoxycyclohex-2-en-1-one |
Specifications
5323-87-5 | |
76°C to 78°C (1 mmHg) | |
C8H12O2 | |
MFCD00001580 | |
3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone | |
JWCFJPLIRVYENQ-UHFFFAOYSA-N | |
3-ethoxycyclohex-2-en-1-one | |
79216 | |
99% |
1.04 | |
107°C (224°F) | |
1.506 | |
2042049 | |
Slightly soluble in water. | |
CCOC1=CC(=O)CCC1 | |
140.18 | |
140.18 | |
3-Ethoxy-2-cyclohexen-1-one |
Safety and Handling
missing translation for 'einecsNumber' : 226-190-7
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.