missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylamino-1,2,4-dithiazole-5-thione, 97%, Thermo Scientific™

Product Code. 11325319
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Dimethylamino-1,2,4-dithiazole-5-thione, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29220-04-0
Molecular Formula C4H6N2S3
Molecular Weight (g/mol) 178.286
MDL Number MFCD00052567
InChI Key IDRNGWAVAQNFSE-UHFFFAOYSA-N
Synonym 3-dimethylamino-1,2,4-dithiazole-5-thione, 5-dimethylamino-3h-1,2,4-dithiazole-3-thione, 5-dimethylamino-1,2,4-dithiazole-3-thione, bio9c6, acmc-20an52, 5-dimethylamino-1,2,4-dithiazoline-3-thione, 3h-1,2,4-dithiazole-3-thione, dimethylamino
PubChem CID 2747561
IUPAC Name 5-(dimethylamino)-1,2,4-dithiazole-3-thione
SMILES CN(C)C1=NC(=S)SS1

Specifications

Melting Point 175°C to 177°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 178.31
Percent Purity 97%
Chemical Name or Material 3-Dimethylamino-1,2,4-dithiazole-5-thione
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.