missing translation for 'onlineSavingsMsg'
Learn More

3'-Deoxyadenosine, 98%, Thermo Scientific Chemicals

Product Code. 15446859
Click to view available options
Quantity:
10 mg
25 mg
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-03-0
Molecular Formula C10H13N5O3
Molecular Weight (g/mol) 251.25
MDL Number MFCD00037998
InChI Key OFEZSBMBBKLLBJ-PNFQIUPPNA-N
Synonym cordycepin, 3'-deoxyadenosine, cordycepine, 9-cordyceposidoadenine, adenosine, 3'-deoxy, 9-beta-d-3'-deoxyribofuranosyl adenine, unii-gz8vf4m2j8, 9h-purine, 6-amino-9-3-deoxy-beta-d-ribofuranosyl, ccris 3233, gz8vf4m2j8
PubChem CID 6303
ChEBI CHEBI:29014
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)C[C@H]3O)C2=NC=N1

Specifications

Color White
Quantity 10 mg
UN Number UN2811
Beilstein 35194
Merck Index 14,2529
IUPAC Name (2R,3R,5S)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolan-3-ol
Formula Weight 251.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3'-Deoxyadenosine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'